Technical Resources - Media Formulations
NZY Broth, powder
INTENDED USE: Maintenance and propagation of bacteriophage lambda. INSTRUCTIONS FOR USE: Suspend 21 g in 1 L of demineralized water. Autoclave for 15 min at 121°C. RECOMMENDED STORAGE CONDITION: 15°C TO 30°C.
  • Catalog Number(s)
ComponentsMolecular WeightConcentration (mg/L)mM
Inorganic Salts
Magnesium Chloride (anhydrous)0.94Infinity
Sodium Chloride (NaCl)5.0Infinity
Other Components
Citric acid anydrous0.19Infinity
SELECT Peptone 140 (pancreatic digest of casein)9.8Infinity
SELECT Yeast Extract autolyzed low sodium5.0Infinity
Reference:
Related Products: