Technical Resources - Media Formulations
21600 - DPBS, powder, no calcium, no magnesium
ComponentsMolecular WeightConcentration (mg/L)mM
Inorganic Salts
Potassium Chloride (KCl)75.0200.02.6666667
Potassium Phosphate monobasic (KH2PO4)136.0200.01.4705882
Sodium Chloride (NaCl)58.08000.0137.93103
Sodium Phosphate dibasic (Na2HPO4) anhydrous142.01150.08.098592
Reference:
1. Dulbecco, R. and Vogt, M., (1954) Plaque formation and isolation of pure lines with Poliomyelitis viruses. J. Exp. Med., 98:167.
Related Products:
14190 - DPBS, no calcium, no magnesium
Cat No: 14190086 , 14190094 , 14190144 , 14190169 , 14190185 , 14190235 , 14190250 , 14190342 , 14190359 , 14190367
14200 - DPBS, 10x, no calcium, no magnesium
Cat No: 14200059 , 14200067 , 14200075 , 14200083 , 14200091 , 14200166