Technical Resources - Media Formulations
14155 - EBSS, no calcium, no magnesium, no phenol red
  • Catalog Number(s)
ComponentsMolecular WeightConcentration (mg/L)mM
Inorganic Salts
Potassium Chloride (KCl)75.0400.05.3333335
Sodium Bicarbonate (NaHCO3)84.02200.026.190475
Sodium Chloride (NaCl)58.06800.0117.24138
Sodium Phosphate monobasic (NaH2PO4-H2O)138.0140.01.0144928
Other Components
D-Glucose (Dextrose)180.01000.05.5555553
Referencia
1. J. Cancer Inst. (1943) 4, 167.