Processing time and protein recovery using 20-100 mL protein concentrators
90 mL volumes of four different protein solutions were concentrated over 30-fold in a swinging bucket rotor at 1200 x g at 25°C, using 5K, 10K, 30K, or 100K MWCO Pierce Protein Concentrators PES (20-100 mL).
90 mL volumes of four different protein solutions were concentrated over 30-fold in a swinging bucket rotor at 1200 x g at 25°C, using 5K, 10K, 30K, or 100K MWCO Pierce Protein Concentrators PES (20-100 mL).
Related Products
Related Images