MDM2 Antibody (MA1113) in WB
Western blot analysis of MDM2 was performed with 10 µg of A549 cells transfected with Transfection Reagent alone (Lane 1), 100nM Non-Targeting control siRNA (Lane 2), or 100nM siRNA against MDM2 (Lane 3). Proteins were resolved using a NuPAGE® Novex 4-12% Bis-Tris Gel (NP0322BOX), XCell SureLock™ Electrophoresis System (EI0002), and a protein size ladder. Proteins were wet transferred to a Pierce Nitrocellulose Membrane (Product # 88025) OR Pierce PVDF Membrane (Product # 88518) and blocked with Pierce Starting Block T20 (PBS) Blocking Buffer (37539) for 1 hour at room temperature. MDM2 was detected at ~ 74 kDa using MDM2 Mouse monoclonal antibody (Product # MA1-113) diluted in Pierce Starting Block T20 (PBS) Blocking Buffer 4°C overnight on a rocking platform. Pierce Goat Anti-Mouse (product # 031437) HRP-Conjugated Antibodies at a 1:2500 dilution were used and chemiluminescent detection was performed using Pierce Supersignal West Dura Maximum Sensitivity Substrate (Product # 37071). Relative density of the bands normalized to GAPDH (36 kDa). MDM2 Antibody (Product # MA1-113) confirms silencing of MDM2 expression.
Western blot analysis of MDM2 was performed with 10 µg of A549 cells transfected with Transfection Reagent alone (Lane 1), 100nM Non-Targeting control siRNA (Lane 2), or 100nM siRNA against MDM2 (Lane 3). Proteins were resolved using a NuPAGE® Novex 4-12% Bis-Tris Gel (NP0322BOX), XCell SureLock™ Electrophoresis System (EI0002), and a protein size ladder. Proteins were wet transferred to a Pierce Nitrocellulose Membrane (Product # 88025) OR Pierce PVDF Membrane (Product # 88518) and blocked with Pierce Starting Block T20 (PBS) Blocking Buffer (37539) for 1 hour at room temperature. MDM2 was detected at ~ 74 kDa using MDM2 Mouse monoclonal antibody (Product # MA1-113) diluted in Pierce Starting Block T20 (PBS) Blocking Buffer 4°C overnight on a rocking platform. Pierce Goat Anti-Mouse (product # 031437) HRP-Conjugated Antibodies at a 1:2500 dilution were used and chemiluminescent detection was performed using Pierce Supersignal West Dura Maximum Sensitivity Substrate (Product # 37071). Relative density of the bands normalized to GAPDH (36 kDa). MDM2 Antibody (Product # MA1-113) confirms silencing of MDM2 expression.
Related Products
Related Images