Search
BPAE cells were fixed, permeabilized, and blocked using the Image-iT® Fixation/Permeabilization Kit (R37602). Samples were then incubated without (A) or with (B) Image-iT® FX Signal Enhancer ReadyProbes® reagent (R37107). Peroxisomes were labeled using an anti-PMP70 antibody (718300) followed by detection with an Alexa Fluor® 594 secondary (A11005). Actin was stained using ActinGreen™ (R37110) and nuclei were stained with NucBlue® Fixed (R37606).