iodoTMT Reagents
(A) iodoTMTzero reagent showing key structural features, including MS/MS fragmentation sites by higher energy collision dissociation (HCD) and electron transfer dissociation (ETD). (B) Structures of iodoTMTsixplex reagents showing the mass (Da) of each reporter and the distribution of normalizing isotopes.
(A) iodoTMTzero reagent showing key structural features, including MS/MS fragmentation sites by higher energy collision dissociation (HCD) and electron transfer dissociation (ETD). (B) Structures of iodoTMTsixplex reagents showing the mass (Da) of each reporter and the distribution of normalizing isotopes.
Related Products
Related Images