Affinity determination of 3 different lots of Goat anti-Rabbit IgG (H+L) Superclonal™ Secondary Antibody, Unconjugated
Histogram showing the affinity (Kd) of Goat anti-Rabbit IgG (H+L) Superclonal™ Secondary Antibody, Unconjugated (A27033) to Rabbit IgG. Based on kinetic characterization of Superclonal™ antibody using the Octet system (forte Bio) the affinity of three lots of the Superclonal™ antibody was consistently in the low picomolar (pM) range.
Histogram showing the affinity (Kd) of Goat anti-Rabbit IgG (H+L) Superclonal™ Secondary Antibody, Unconjugated (A27033) to Rabbit IgG. Based on kinetic characterization of Superclonal™ antibody using the Octet system (forte Bio) the affinity of three lots of the Superclonal™ antibody was consistently in the low picomolar (pM) range.
Related Products
Related Images