Search
Stripping and reprobing blots for similar molecular weight targets with Restore Western Blot Stripping Buffer. A431 cell lysate was diluted to 125µg/mL in electrophoresis reducing sample buffer and 1:1 serial dilutions were made. 10µL of each dilution (1250ng to 39ng of total protein) were separated by SDS-PAGE and the protein transferred to 0.45µm nitrocellulose membranes (Part No. 88018). The membrane was blocked with 5% non-fat dry milk in 1X PBS Tween-20 Buffer (Part No. 28352) and analyzed by Western blot using SuperSignal West Dura Extended Duration Substrate (Part No. 34076) and the myECL Imager (3×3 binning) (Part No. 62236). The first target (panel 1) was detected by probing with Anti-PDI Monoclonal Antibody (Part No. MA3-019) at 0.33µg/mL followed by Goat anti-Mouse Horseradish Peroxidase Conjugate (Part No. 31460) at 6.7ng/mL and imaged. Next, the blot was stripped in Restore Western Blot Stripping Buffer (Part No. 21059) for 15 minutes at 37°C, washed in 1X PBS Tween-20, incubated with substrate and imaged to check for stripping efficiency (panel 2). The second target (panel 3) was detected by reblocking the membrane and probing with Anti-Actin Monoclonal Antibody (Part No. MA1-744) at 0.5µg/mL followed by the anti-mouse HRP conjugate at 6.7ng/mL and imaged. The blot was stripped again (panel 4) and then probed for multiple targets (alpha-Tubulin, Part No. 62204 at 0.2µg/mL; PDI as above; and Hsp90, Part No. PA3-013 at 0.14µg/mL), and imaged as described.