Phospho-Tuberin / TSC2 pSer939 Antibody (710395) in FACS
Flow cytometry analysis of Phospho-Tuberin pSer939 in HepG2 cells using a Phospho-Tuberin pSer939 recombinant rabbit oligoclonal antibody (Product # 710395). Cells were fixed and permeabilized using FIX & PERM (Product # GAS004) reagent, and detection was performed using an Alexa Fluor 488 goat anti-rabbit IgG (right peak) compared to an isotype control (left peak).
Flow cytometry analysis of Phospho-Tuberin pSer939 in HepG2 cells using a Phospho-Tuberin pSer939 recombinant rabbit oligoclonal antibody (Product # 710395). Cells were fixed and permeabilized using FIX & PERM (Product # GAS004) reagent, and detection was performed using an Alexa Fluor 488 goat anti-rabbit IgG (right peak) compared to an isotype control (left peak).
Related Products
Related Images