May 21, 2019
- May 23, 2019
2019 MAM Symposia
Confident, reproducible, and fast characterization of therapeutic proteins is critical for drug product quality and patient safety. The Multi-Attribute Method (MAM) is a mass spectrometry-based peptide mapping & monitoring workflow used to quantify a range of product quality attributes and detect new impurities simultaneously, in a single analytical protocol.
Attend a seminar to connect with industry experts in the Multi-Attribute Method from Symphogen; NIBRT; Sanofi; Novo Nordisk; and Thermo Fisher Scientific, network with users, and learn innovations in the workflow.
- Learn how peptide monitoring using high resolution accurate mass (HRAM) mass spectrometry provides the necessary resolution of peptides and their modifications to afford confident measurement of potential critical quality attributes (CQAs)
- See how compliant-ready software delivers the necessary new peak detection to allow for flagging of untargeted differences
- Discover how MAM can be used from discovery through to manufacturing
May 21, 2019 - May 21, 2019
9:00 – 14:30
Pullman Europe Hotel
Clarastrasse 43
Basel, Switzerland 4058
Seminar topics will include:
- Introduction to the BioPharmaceutical MAM Workflow
- Sample Preparation & Column Choice for MAM
- MAM Software Overview - Identifying & Monitoring CQAs
- Installation, Qualification, Training and Support for MAM
May 22, 2019 - May 22, 2019
9:00 – 14:30
Symphogen A/S
Pederstrupvej 93
Ballerup, Denmark 2750
Seminar topics will include:
- Introduction to the BioPharmaceutical MAM Workflow
- Sample Preparation & Column Choice for MAM
- MAM Software Overview - Identifying & Monitoring CQAs
- Installation, Qualification, Training and Support for MAM
May 23, 2019 - May 23, 2019
12:00 – 17:00
Milton Park
Park Dr
Abingdon, UK OX14 4RT
Seminar topics will include:
- Introduction to the BioPharmaceutical MAM Workflow
- Sample Preparation & Column Choice for MAM
- MAM Software Overview - Identifying & Monitoring CQAs
- Installation, Qualification, Training and Support for MAM