Thermo Fisher Scientific Product Spotlights at European Microscopy Congress 2024
Product spotlights
Visit our booth to learn more about our wide range of instruments and software.
Introducing our new EELS-TEM: The first fully integrated advanced analytical (S)TEM
New SEM with EDS and EBSD integration. Scanning electron microscope for imaging, chemical and structural analysis. The new SEM is the ultimate tool for fully integrated high resolution SEM imaging with integrated Chemical (EDS) and Structural (EBSD) analysis.
Amira Software
Thermo Scientific Amira Software is a powerful 2D–5D solution that delivers a comprehensive range of data visualization, processing, and analysis capabilities to help life sciences and biomedical researchers gain invaluable insights into their data.                                                                                                                                       
Avizo Software
Thermo Scientific Avizo Software provides optimized workflows for advanced materials characterization and quality control from a single environment, helping you analyze and visualize imaging data to better understand the structure, properties, and performance of your materials.
CleanMill Broad Ion Beam System
The Thermo Scientific CleanMill Broad Ion Beam System is a complete ion beam polishing solution for SEM applications in materials science, enabling optimal imaging and analysis of materials that require a pristine surface, including beam- and air-sensitive materials.
Connected Care Services
Connected Care Services maximize your electron microscope’s uptime to improve the reliability of your workflow, optimize system utilization, and achieve faster time to data.                                                                                                                                                                                                                                       
Thermo Scientific CryoFlow Software
Thermo Scientific CryoFlow Software is a paperless data management solution that optimizes cryo-electron microscopy (cryo-EM) workflows for single particle analysis (SPA) and cryo-electron tomography (cryo-ET).                                                                                              
Glacios 2 Cryo-TEM
The Thermo Scientific Glacios 2 Cryo-Transmission Electron Microscope delivers higher throughput, enhanced image quality, and simpler operation than the previous generation, helping you easily collect near-atomic data from a broad range of biological targets. High-quality data collection is possible thanks to hardware improvements such as E-CFEG and the integration of innovative Thermo Scientific Smart EPU Software.                                                                                                                                
Helios 5 Hydra DualBeam PFIB-SEM
The Thermo Scientific Helios 5 Hydra DualBeam PFIB-SEM can deliver argon, nitrogen, oxygen, and xenon as the primary beam and switch between species in under 10 minutes, making it easy to select the ions best suited to your samples and use cases.                                                                              
Hydra Bio Plasma-FIB
The Thermo Scientific Hydra Bio Plasma-FIB offers versatility in volume electron microscopy and cellular cryo-tomography at varying temperatures. It supports a range of ion species for optimal sample preparation with standard embedding media and techniques. Its applications include high-resolution and large-area serial PFIB imaging, array tomography, and correlative microscopy, streamlining the preparation process for cryo-tomography.
Krios G4 Cryo-TEM
The award-winning Thermo Scientific Krios G4 Cryo-Transmission Electron Microscope helps you unravel life at the atomic level — easier, faster, and more reliably than ever before. The most compact TEM in its class, the Krios G4 Cryo-TEM consists of a highly stable 300 kV TEM platform and the industry-leading Autoloader (a cryogenic sample manipulation robot), making it ideally suited for automated applications such as cryo-electron tomography, single-particle analysis, and micro-electron diffraction.   
Nicolet RaptIR+ FTIR Microscope with Nicolet iS50 FTIR Spectrometer
The research-grade Thermo Scientific Nicolet RaptIR+ Microscope can help you rapidly locate and identify trace materials, inclusions, impurities, and microparticles as well as determine the distribution of materials within a sample with incredible clarity and precision. 
Phenom ParticleX Desktop SEM 
The Thermo Scientific Phenom ParticleX Desktop SEM is a dedicated and automated particle analysis solution for failure analysis, quality control and process improvement in a variety of industries. Powered by Thermo Scientific Perception Software, it delivers advanced automated particle analysis, classification, and unparalleled reporting. 
Phenom Pharos G2 FEG-SEM
The Thermo Scientific Phenom Pharos G2 FEG-SEM brings field emission SEM to your tabletop, offering easy operation, fast sample exchange, and powerful performance to help academic and industrial laboratories quickly complete imaging and analysis jobs.  
Smart EPU Software
Thermo Scientific Smart EPU Software uses AI to collect and process cryo-EM single particle analysis data, helping to improve throughput, predictability, and reproducibility with less hands-on time no matter your level of experience.                                                                                                                              
Spectra Ultra Scanning Transmission Electron Microscope
The Thermo Scientific Spectra Ultra Scanning Transmission Electron Microscope enables ultra-fast EDX mapping, tomography, ultra-low dose EDX spectroscopy, and live in situ EDX while optimizing conditions to mitigate beam damage and improve EDX count rate.
Talos (S)TEMs with Cold FEG
The Thermo Scientific Talos F200X G2 (S)TEM combines high-resolution S/TEM and TEM imaging with industry-leading energy dispersive X-ray spectroscopy. It can now be powered by a new ultra-high-brightness cold field emission gun (X-CFEG), a small source size (tip), and low energy spread for faster and higher-resolution imaging.                                                                                                                          
Tomo Live Software 
Thermo Scientific Tomo Live Software is an optional add-on for Tomography 5 Software that offers automatic, on-the-fly reconstruction with data curation for review in any browser, anytime, anywhere.                                                                                                                       
For Research Use Only. Not for use in diagnostic procedures.