Search Thermo Fisher Scientific
If you have questions about these webinar sessions or regarding your registration please email analyze.eu@thermofisher.com
You cannot attend the live sessions? Please register anyway. You will automatically receive the link to the webinar recording the day after broadcast.
All webinars listed below are in German language.
These webinar recordings are instantly available. View them any time!
Please note that for most webinars below you could use the question&answer chat also when viewing the webinar recordings. Our experts will get back to you within a few days up to a week.
Routine testing laboratories often run multiple wet chemical tests per sample. They typically split the sample into portions depending on the analytes run them using different wet chemical instruments, such as ion meters, auto titrators, spectrophotometers, flow injection analyzers, and more. These instruments themselves are often automated and deliver precise and accurate results, but overall analysis of a samples for multiple parameters takes long and have limited options for automation. Why operate with several analytical instruments, when you have a solution to run your sample for multiple parameters on one system only? This webinar covers how discrete industrial analyzers (DIA) offer automation of up to 350 tests per hour. Our DIA application experts discuss how these systems are an efficient alternative to other techniques like traditional flow analyzers. The unique low volume cuvette design produces cost saving in reagents and waste.
View these sessions now to hear about the industry´s most liberating technology delivering reliable results for multiple parameters - from a single sample.
Thermo Scientific Gallery, Gallery Plus and Gallery Plus Beermaster discrete industrial analyzers
Learn about discrete industrial analyzers in the food and beverages testing laboratory and how it can be applied for sugars, total proteins, amino acids, iron, copper and total polyphenol analysis; plus specific wine solutions including total sulfite, glycerol and tartaric acid, beer and malt applications such as bitterness and Sulphur dioxide.
Learn about discrete industrial analyzers in the environmental testing laboratory and how it can be applied for analysis such as alkalinity, conductivity, nitrite, urea, total oxidized nitrogen (TON), silica and total hardness in a variety of sea, natural and waste waters, and soil testing.
Discover how the new Thermo Scientific Exploris Orbitrap GC-MS is opening new opportunities for analytical testing and research discovery. This presentation also includes an online video demonstration of the system by the application chemists from our lab in Dreieich.
Register for the Orbitrap GC-MS webinar in German language »
Do you want to know why so many users doing trace elemental analysis choose Thermo Scientific iCAP PRO ICP-MS systems?
View this recent webinar recording and discover an ICP-OES that offers long-term stability, low detection limits and more accurate results you want and need.
Topics:
HPLC combined with this universal detection technology overcomes many analytical challenges that cannot be solved with conventional detectors. Newest technical improvements in the Thermo ScientificCharged Aerosol Detectors show high sensitivity, linearity and flexibility with high user-friendliness.
Solvent free sample preparation - Why headspace as a good sampling technique!
In this webinar we focus on sample preparation for gas chromatography. The following topics are covered:
Speakers:
Thomas Stegemann, Sales Support Specialist and Dr. Klaus Schrickel, Senier Sales Support Specialist
Thermo Fisher Scientific, Dreieich
Routine HPLC laboratories, especially pharmaceutical QA/QC laboratories, have grown resigned to the need for intensive resources dedicated to HPLC system and data monitoring to ensure delivery of results.
Simple to the Core. Eliminate disruptions to your routine analytical work with the Thermo Scientific Vanquish Core HPLC systems. No matter which systems you currently use, switching to Vanquish Core is simple and seamless.
New standards in productivity. As a member of the Vanquish product line, the Vanquish Core HPLC systems share the Vanquish values of hardware precision, detector sensitivity, and simplicity of operation. Vanquish Core HPLC systems deliver exactly what you need for your routine analyses.
Continuous, worry-free operation. The Thermo Scientific Vanquish Solvent Monitor delivers uninterrupted sequence runs through the avoidance of dry solvent channels and supports your laboratory’s compliance efforts through automatically logging eluent changes. By monitoring waste container levels, you can ensure your laboratory’s smooth operation by preventing hazardous spills.
Discover how to maximize lab productivity through new dedicated tools, minimize instrument downtime using teh new Vanquish Solvent Monitor and learn how we have helped you simplify regulatory compliance.
Discrete industrial analyzers (DIA) provide rapid and automated photometric (Colorimetric and enzymatic) and electrochemical (pH and conductivity) analysis giving fast, reproducible results. With discrete cell technology, multiple analytes can be run simultanously reducing operation and run time.
This webinar will cover how discrete industrial analyzers work and introduce the Thermo Scientific™ Gallery™, Gallery™ Plus and Gallery™ Plus Beermaster systems, with automation up to 350 tests per hour. Our DIA specialist will also discuss how these systems are an efficient alternative to traditional flow analyzers and how the unique low volumne cuvette design produces cost savings in reagents and waste. Finally we show how this technology can be applied in a food and beverage testing laboratory for test such as sugars, total protein, amino acids, iron, copper and total polyphenol. Plus specific wine solutions including total sulfite, glycerol and tartaric acid and beer and malt applications such as bitterness and sulphur dioxide.
Speaker:
Dr. Marco Rastetter, Senior Sales Support Specialist
Thermo Fisher Scientific, Dreieich
Discrete industrial analyzers (DIA) provide rapid and automated photometric (Colorimetric and enzymatic) and electrochemical (pH and conductivity) analysis giving fast, reproducible results. With discrete cell technology, multiple analytes can run simultanously reducing operating and run time.
This webinar will cover how discrete industrial analyzers work and introduce the Thermo Scientific™ Gallery™ and Gallery™ Plus systems, with automation enabling up to 350 tests per hour. Our DIA specialist will discuss how this technique is an effcient alternative to traditional flow analyzers and how the unique low-volumne cuvette design produces cost savings in reagents and waste.
Finally we show how this technology can be applied in an environmental laboratory for test such as alkalinity, conductivity, nitrite, urea, total oxidized nitrogen (TON), silica and total hardness in a variety of sea, natural and wast waters and soil testing.
Speaker:
Dr. Marco Rastetter, Senior Sales Support Specialist
Thermo Fisher Scientific, Dreieich
In this webinar we will introduce you to the new Thermo Scientific iCAP PRO ICP-OES, a fast, powerful and easy-to-use system with which you can achieve the lowest detection limits even in highly concentrated samples.
The 2020 virtual Swiss HPLC user meeting
Day one topics include:
Day two topics:
From the Sample to the result - The journey of a molecule from sample to detection and which steps it takes along that way
In this webinars we present from the perspective of a molecule all relevant steps of the sampling process, storage, preparation, up to detection by a GC-MS. The molecule looks at every step on this way with a certain sense of humour.
This presentation is for everyone in the lab dealing with sampling, sample storage and preparation, up to detection.
After the presentation we will have time to answer your questions.
This webinar will provide an overview on new ways to improve your detection limits and avoid costly re-testing.
A new generation of ICP-MS instruments is entering routine analysis. While established instruments with single quadrupole technology are reaching their limits for some elements (e.g. phosphorus, arsenic or selenium) despite significant progress, instruments with triple quadrupole technology can resolve a wide variety of interferences, as well as significantly increase sensitivity in some cases. This enables improvement in the quality of data obtained, as well as an improvement in the laboratory procedures, as the need of re-testing of possible incorrect samples can be avoided. In particular laboratories running environmental and food analysis will benefit from this, but also other areas, such as continuous quality control in the metal processing industry or material science.
Despite the many options to separate potential interferences by using reactive gases such as oxygen and ammonia, modern triple quadrupole instruments are easy to operate thanks to innovative software tools - even for unexperienced users.
Keep your lab up and running 24 hours a day, 7 days a week
A business continuity plan (BCP) is an essential part of any business strategy to ensure continuous, uninterrupted lab operations regardless of internal or external events. As your reliance on digital data systems and recent cyberattacks has grown, it has never been more important than it is today to ensure that your lab data systems are protected from the effects of such attacks.
In this webinar, we discuss how choosing your chromatography data system (CDS) can help you plan unforeseen events effectively and thus maintain the 24/7 operation of your lab.
We provide an example of how hardware and software can work together to maintain and secure business continuity.
In this webinar we demonstrate applications for rapid UHPLC targeted screening for compounds such as food additives, food dyes and endogenous food compounds such as catechins, anthocyanins and steviol glycosides. This webinar focuses on utilizing the capabilities of both instrument and alternative column chemistries to maximise method reproducibility, sensitivity and efficiency of separations.
The Thermo Scientific Vanquish UHPLC platform was introduced five year ago in 2014 and revolutionized UHPLC through innovative design to deliver the requirements demanded by the modern chromatographer. During this webinarwe demonstrate how this LC platform has delivered these features in to tangible user benefits and how the revolution in liquid separations through Vanquish has progressed over the past five years, with real-life examples, cumulating in the recent Vanquish Duo systems. This dual LC system allows three workflows (Dual LC, Tandem LC or LC-MS and Inverse Gradient) and two independent flow paths in one complete system solution.
Ever considered which reagents and grades are optimum for your separations? How best to keep your baselines low? In this webinar you will learn about the different grades of reagents, recommendations for their use based on application, and materials and processes to clean and maintain your instruments.
Whether volatile or stable, highly automated sampling solutions are a must for increased productivity in routine labs. New technological developments provide largely automated workflows and the reliability required for continuous operation and minimal downtime. This webinar introduces and explains the applicability of highly automated sample preparation for gas chromatography. Learn how innovation and automation help you save time and money in your daily workflow.
The screening and quantification of veterinary medicinal products in food is a demanding but essential application component for food safety. In this webinar, learn how the holistic, robust and multi-stage LC-MS/MS method for veterinary medicinal products can be applied to a variety of matrices such as beef, salmon fillet and milk.