Live Demos of Thermo Fisher Scientific Solutions at International Microscopy Congress 2023
Thermo Scientific instrument presentations at IMC20
We are excited to provide live demonstrations of the following products. If you are interested in seeing something in particular in action, please register in advance for a demo slot.
Apreo 2 SEM
The Thermo Scientific Apreo 2 FEG-SEM delivers outstanding image quality with high contrast even for beam-sensitive materials. Combined with Thermo Scientific ChemiSEM Tehcnology, the system delivers simultaneous access to compositional information via the intuitive user interface. This eliminates typical steps in the workflow resulting in the shortest time to reliable results possible.
The Thermo Scientific Helios 5 Hydra PFIB-SEM can deliver argon, nitrogen, oxygen, and xenon as the primary beam and switches between species in under 10 minutes, making it easy to select the ions best suited to your samples and use cases.
Thermo Scientific Talos F200i and Talos F200X G2 S/TEMs can now be powered by a new ultra-high-brightness cold field emission gun (X-CFEG) with high brightness, a small source size (tip), and low energy spread for faster, higher-resolution imaging.
The Thermo Scientific Spectra Ultra Scanning Transmission Electron Microscope enables ultra-fast EDX mapping, tomography, ultra-low dose EDX spectroscopy, and live in situ EDX while optimizing conditions to mitigate beam damage and improve EDX count rate.
The new Thermo Scientific Glacios 2 Cryo-Transmission Electron Microscope delivers higher throughput, enhanced image quality, and simpler operation than the previous generation, helping you easily collect near-atomic data from a broad range of biological targets.
The Thermo Scientific Tundra Cryo-TEM, in combination with the Thermo Scientific Falcon C Detector, allows structural analyses on challenging proteins (<100 kD) with unprecedented ease of use. Easily load and unload cryo-EM grids in a contamination free state, and screen room temperature negative stain grids and plastic sections.
The Thermo Scientific™ Hydra Bio™ Plasma-FIB is designed for volumetric imaging of frozen-hydrated and plastic-embedded biological samples. The Hydra Bio Plasma-FIB also features proven automation and cryo-technologies for versatile cryo-electron tomography lamella preparation.
Thermo Scientific Phenom ParticleX Desktop SEMs deliver a comprehensive plug-and-play SEM-EDS software solution for particle analysis workflows. Featuring automated characterization, verification, and classification of materials, the instruments support your production with fast, accurate, and reliable data that adheres to industry regulations and standards.
Register on-site at Thermo Fisher booth #S01
Axia ChemiSEM with Perception Software
The Thermo Scientific Axia ChemiSEM Scanning Electron Microscope is a next-generation, alignment-free SEM platform with unique, live, quantitative energy-dispersive X-ray spectroscopy (EDS) mapping, making it the instrument of choice for rapid analysis — even for users who are new to SEM. Thermo Scientific Perception Software (already available on our desktop SEM systems) brings automated particle and inclusion analysis to the system. Running unattended overnight, it collects imaging and EDS data of the particles and turns data into insights with application-specific reporting.