Ruben Pawellek - Institut für Pharmazie und Lebensmittelchemie, University of Würzburg
Abstract:
During the last decade, CAD has become an established detection technique in the field of pharmaceutical analysis. Recently, the European Pharmacopoeia introduced the detector for the analysis of weakly chromophoric drugs. This presentation provides an overview of recent CAD applications for compendial quality control purposes focusing on the impurity profiling of drug substances. In this regard, the in-line coupling of UV-CAD offers numerous advantages when impurities with opposing physico-chemical properties are analysed simultaneously, e.g. volatile impurities and impurities with weak chromophores.1
The application of the CAD in a regulated environment requires careful selection of the detector settings to comply with the demands for robust and sensitive methods.2 Here, we present optimization strategies for the PFV setting of the CAD that can help to linearize the detector signal in a manner conforming to GMP regulations.3 Moreover, we include a brief discussion about pros and cons of IPC and HILIC for impurity profiling focusing on the impact of the respective separation technique on CAD sensitivity.
1 R. Pawellek, K. Schilling, U. Holzgrabe, Impurity profiling of l-aspartic acid and glycine using high-performance liquid chromatography coupled with charged aerosol and ultraviolet detection, J. Pharm. Biomed. Anal. 183 (2020) 113149.
2 K. Schilling, R. Pawellek, K. Lovejoy, T. Muellner, U. Holzgrabe, Influence of charged aerosol detector instrument settings on the ultra-high-performance liquid chromatography analysis of fatty acids in polysorbate 80, J. Chromatogr. A 1576 (2018) 58-66.
3 R. Pawellek, T. Muellner, P. Gamache, U. Holzgrabe, Power function setting in charged aerosol detection for the linearization of detector response – optimization strategies and their application, J. Chromatogr. A 1637 (2021) 461844.
Learning points
- CAD is essential for the impurity profiling of weakly chromophoric drugs
- Optimization of CAD settings is crucial for the development of robust and reproducible methods for a compendial application
- UV-CAD can be easily applied to broaden the detection scope
System used - Thermo Scientific Vanquish Flex Binary UHPLC System with Vanquish Charged Aerosol Detector H
Application - Pharmaceutical analysis
Register now!