Tradeshow
Aug 22, 2021 - Aug 26, 2021
Online
Microscopy Congress 2021
If you are a materials scientist, our electron microscopy instruments and workflow solutions provide you the ability to view things at the nanoscale, helping you develop better batteries, safer materials used in planes, or even develop drug delivery mechanisms.
If you are a life sciences researcher, our electron microscopes and software solutions offer a pathway to a better understanding of how diseases and viruses work so you can develop treatments. Using cryo-EM, you can unravel 3D structures of proteins not easily crystalized for X-ray diffraction (XRD) or used for nuclear magnetic resonance (NMR) experiments.
See some of the solutions we shared at MC2021 below.
Meet the products for Materials Science
Find out how to get much better statistics on nanoparticles or precipitates in a much faster way by using the unique APW pack that enables fully automated unattended runs from acquisition to processing. It also significantly improves the repeatability since there is no operator bias. Using this pack enables you to optimize your product in a much faster way. 
Consistency, reliability, and integrity can be increased with automation. This session presents the advanced automation capabilities of the Thermo Scientific Metrios (S)TEM for semiconductor analysis.
In this presentation on our Thermo Fisher Scientific Spectra Ultra (S)TEM you will learn about the highest resolution S/TEM platform for the characterization of the widest range of materials. The Spectra Ultra (S)TEM builds on the already high-performing Spectra 300 (S)TEM with new EDX detector technology and rapid accelerating voltage switching (30–300 kV) to accommodate more complex and beam-sensitive specimens.
In this session, we will show you how the Helios 5 Laser PFIB enables:
  • Fast, millimeter-scale characterization with nm resolution using a fully integrated femtosecond laser
  • Optimized milling for a wide range of materials due to fast ion switching capability (Xe/Ar/N/O)
  • Automated, in situ, Ga-free TEM sample preparation using Thermo Scientific AutoTEM 5 Software
The Thermo Scientific Phenom Pharos G2 Desktop FEG-SEM brings field emission SEM to your tabletop. View this Meet the Product session to learn how it makes FEG performance accessible to a wide range of users thanks to its attractive form factor and short training required.
The new Thermo Scientific Apreo 2 SEM expands access to high-performance imaging and analytics to all levels of microscopy expertise. With Thermo Scientific ChemiSEM Technology, a unique live elemental imaging capability, compositional information is always available, through the intuitive interface. View this Meet the Product session and learn more about this scanning electron microscope for versatile, high-performance materials imaging and analysis.
Meet the products for Life Sciences
Learn how imaging data analytics and management software help life and materials scientists tackle today’s and tomorrow’s health and environmental challenges.
At Thermo Fisher Scientific, our goal is to give you all the tools and resources you need to be successful with your scientific instrument. Our secure, web-based Connected Care portal layers system data with advanced algorithms to deliver impactful insights about your system’s productivity.
  • Track system utilization and output, to ensure you’re getting the most value from your instrument.
  • View real-time system health status to ensure your system is primed for your next sample run.
  • Conveniently access service history, reports, and applications credits.
Meet the experts - Materials Science
Meet the Phenom Pharos G2 Desktop SEM – Intuitive evidence for your research
Do you need the performance of a field emission (FEG) SEM, but also have limited space, no time to wait for sample loading, and no capacity to train all users? Watch the Thermo Scientific Phenom Pharos G2 Desktop FEG-SEM demonstration to:
  • Learn how FEG-level performance has become more accessible than ever
  • Discover the amazing image quality that a tabletop SEM can deliver
  • See how samples are loaded in just 30 seconds
Automation on the Talos (S)TEM
Find out how automation on the Thermo Scientific Talos (S)TEM delivers better results faster for all users. It significantly reduces the tuning overhead, training is much faster, experiment repeatability improves, and you will be able to complete even more samples and applications per day. Additionally, the system can be used overnight or over the weekend in an automated way.
Meet the Apreo 2 SEM: Unmatched versatility powered by ChemiSEM Technology
Our Thermo Scientific Apreo 2 SEM demonstration will show you how to deal with real-life challenges surrounding high-performance SEM.
  • Do you have difficult samples to investigate?
  • Are insulating materials making your SEM imaging difficult?
  • Is elemental information important for you? Are you interested in the chemistry of your sample?
  • Do you have multiple and/or new users?
  • Do you need a quick result?
Meet Spectra Ultra (S)TEM and see how to analyze the widest range of materials at the atomic scale
View the Thermo Scientific Spectra Ultra (S)TEM Meet the Expert session to hear from our product expert about how our highest resolution (S)TEM platform can characterize the widest range of materials. The Spectra Ultra (S)TEM builds on the already high-performing Spectra 300 (S)TEM with new EDX detector technology and rapid accelerating voltage switching (30–300 kV) to accommodate more complex and beam-sensitive specimens.
Unique, automated, in situ TEM sample preparation workflow for users of any experience level
Learn how the newest generation of Thermo Scientific DualBeam systems, in combination with Thermo Scientific AutoTEM 5 Software, enable the fully automated process of highest quality lamella preparation, from chunking to in situ lift-out, thinning, and final low-kV polishing. In full automation mode, thinned lamellas could be prepared unattended in as little as 45 minutes. In this video you will discover how AutoTEM 5 Software enables even new users to utilize the top system performance and make high throughput and reproducibility of TEM sample preparation accessible to all users.
Meet the experts - Life Sciences
Explore large multi-channel and time series data with Amira Software and the new Xplore5D extension
This presentation will focus on the visualization and image processing of large 3D image data using the new capabilities of Thermo Scientific Amira Software. The webinar will give you the possibility to learn, through a step-by-step demo, the recently added functions of Amira Software’s new Xplore5D extension. XPlore5D offers the fast and easy exploration of large multi-channel and time series imaging data. We will also highlight and demonstrate Deep Learning-based training and prediction modules.
Tundra Cryo-TEM: Cryo-EM for all
Single particle transmission cryo-electron microscopy (cryo-EM) has become one of the dominating methods for high-resolution structure determination of a wide variety of biological macromolecules to facilitate understanding of their functions, mechanisms of action, and protein ligand/drug interactions. The Thermo Scientific Tundra Cryo-TEM is designed for ease of use for non-experts to bring cryo-EM to all. Learn about sample optimization and the new semi-automated loader.
Scientific presentations