Life Technologies GeneArt® Strings™ Now Available Up to Three Kilobases
Untitled Document
Laboratory Products
Media Contact Information:
Suzanne Clancy, ThermoFisher Scientific
760-602-4545
Suzanne.clancy@thermofisher.com
NEW PRODUCT RELEASE:Life Technologies GeneArt® Strings™Now Available Up to Three Kilobases
Date: March 31, 2014
Product:
Life Technologies GeneArt® Strings™ DNA Fragments
Product Image:
chart
Product Overview:
GeneArt® Strings™ DNA Fragments are custom-made, uncloned, double-stranded linear DNA fragments from 100 base pairs (bp) to 3,000 bp in length, assembled from synthetic oligonucleotides using the same process developed for GeneArt® high-quality gene synthesis. Strings™ Fragments are delivered dried, ready for resuspension and screening to enable the identification of the correct clone. Strings™ Fragments are a fast and smart alternative to get your synthetic gene and clone your expression plasmid.
GeneArt® products are for Research Use; not intended for diagnostic use.
Key Applications:
Cloning for optimal protein expression
Features/Benefits:
  • Advantages of synthetic DNA – design flexibility and sequence optimization for robust and reliable expression – along with quick production times of five to eight business days.
  • Tiered pricing system correlates with increasing size of the ordered fragment.
  • Can be edited, optimized, and ordered via the Life Technologies online order portal for a convenient and user-friendly customer experience.
Web Site:
http://www.lifetechnologies.com/us/en/home/life-science/cloning/gene-synthesis/gene-strings-dna-fragments.html
Company Background:
About Thermo Fisher Scientific
Thermo Fisher Scientific Inc. is the world leader in serving science, with revenues of $17 billion and 50,000 employees in 50 countries. Our mission is to enable our customers to make the world healthier, cleaner and safer. We help our customers accelerate life sciences research, solve complex analytical challenges, improve patient diagnostics and increase laboratory productivity. Our four premier brands – Thermo Scientific, Life Technologies, Fisher Scientific and Unity Lab Services – offer an unmatched combination of innovative technologies, purchasing convenience and comprehensive support. For more information, please visit www.thermofisher.com.