Search
Partner with us to build your product development pipeline, or scale-up your processes with bulk quantities of high-quality detergents. We have been supplying bulk and custom-made reagents to the biotechnology and pharmaceutical industries for over 50 years.
We offer a variety of compounds, devices, and resins to suit any application. Whether you are developing an assay for commercialization, constructing a large screening assay for in-house use, or require high-purity reagents for your manufacturing processes, we can provide consistent quality in milligram to kilogram quantities. Reagents are manufactured under ISO 9001 quality systems.
Contact us to discuss your custom or bulk highly-purified detergent requirements.
Our detergents are available in multiple package formats to best meet your individual needs. We offer several high purity, low peroxide-containing detergents that are packaged under nitrogen gas in clear glass ampules, providing superior convenience, quality, and consistency for all detergent applications.
| Detergent | Description | Aggregation number | Micelle MW | MW | CMC (mM) | CMC % w/v | Cloud point (ºC) | Dialyzable |
|---|---|---|---|---|---|---|---|---|
| Triton X-100 | Nonionic | 140 | 90,000 | 647 | 0.24 | 0.0155 | 64 | No |
| Triton X-114 | Nonionic | — | — | 537 | 0.21 | 0.0113 | 23 | No |
| NP-40 | Nonionic | 149 | 90,000 | 617 | 0.29 | 0.0179 | 80 | No |
| Brij®-35 | Nonionic | 40 | 49,000 | 1,225 | 0.09 | 0.1103 | >100 | No |
| Brij-58 | Nonionic | 70 | 82,000 | 1,120 | 0.077 | 0.0086 | >100 | No |
| Tween®-20 | Nonionic | — | — | 1,228 | 0.06 | 0.0074 | 95 | No |
| Tween-80 | Nonionic | 60 | 76,000 | 1,310 | 0.012 | 0.0016 | — | No |
| Octyl-beta-Glucoside | Nonionic | 27 | 8,000 | 292 | 23-25 | 0.6716-0.7300 | >100 | Yes |
| Octylthio Glucoside | Nonionic | — | — | 308 | 9 | 0.2772 | >100 | Yes |
| SDS | Anionic | 62 | 18,000 | 288 | 6-8 | 0.1728-2304 | >100 | Yes |
| CHAPS | Zwitterionic | 10 | 6,149 | 615 | 8-10 | 0.4920-0.6150 | >100 | Yes |
For Research Use Only. Not for use in diagnostic procedures.