On this page:
Purification with ion exchange chromatography resins
Ion exchange chromatography (IEX) separates biomolecules according to their charge. POROS ion exchange chromatography resins can be used in both capture and polishing steps, spanning from early development to large-scale manufacturing. They enable selective binding across a broad pH and conductivity range. Various anion and cation exchange formats are available, offering options suited to different molecule types and process conditions.
Benefits of ion exchange resins
POROS ion-exchange resins use a rigid backbone with large throughpores allowing for rapid purification of biologics at elevated velocities. They are designed to maintain high dynamic binding capacity across broad pH and conductivity ranges, supporting robust process development across therapeutic modalities.
POROS IEX resins help deliver:
  • Superior resolution
  • High throughput
  • High binding capacity
Explore our ion exchange resins
Explore other chromatography resins
Affinity chromatography resins
Affinity resin allows selective capture with high purity and yield in a single capture step. CaptureSelect technology extends beyond antibodies to bispecifics, fragments, Fc-fusion proteins, recombinant proteins, plasma proteins, mRNA, and viral vectors under mild conditions.
Hydrophobic interaction chromatography resins
Hydrophobic interaction chromatography (HIC) separates biomolecules by hydrophobicity. It is often used as a polishing step, improving purification where charge-based methods may be limited.
Mixed-mode chromatography resins
Mixed-mode chromatography resins can improve resolution leveraging a single ligand with ionic and hydrophobic interactions. Mixed-mode cation exchange resins are commonly applied in flow-through polishing to help reduce aggregates and other impurities while maintaining high product recovery.
Protein A chromatography resins
Protein A chromatography is widely applied in antibody purification. MabCaptureC resin offers high binding capacity, alkaline stability, and scalability while keeping cost of goods in mind, making it suitable from process development to cGMP manufacturing.
Skip the packing. Start purifying.
Ready-to-use prepacked chromatography columns eliminate manual column packing and help simplify scale-up from process development through manufacturing. 
Not sure which resin is right for you?
The Bioprocess Resin Selection Guide is an interactive guide that helps scientists evaluate chromatography resins based on molecule type, application, and scale. Identify products that meet your purification goals and process needs by filtering data on resin chemistry, capacity, and operating conditions.
Frequently asked questions
Ion exchange resins are used to purify biomolecules by separating them based on their surface charge. They are applied in capture and polishing steps for antibodies, plasmid DNA, viral vectors, and recombinant proteins. They can also be used in flow-through mode to help reduce impurities such as DNA, viruses, host cell proteins, and product and process-related impurities.
Ion exchange chromatography is used in the purification of advanced modalities, including viral vectors, plasmid DNA, and vaccines. Depending on the process, it functions in bind/elute mode for product capture or in flow-through mode to reduce impurities and separate product variants.
Anion exchange resins carry positively charged groups and bind negatively charged biomolecules such as DNA, plasmid, or viruses. Cation exchange resins carry negatively charged groups and bind positively charged molecules, such as antibodies or proteins with a high isoelectric point.
Resin choice depends on the molecule’s net charge at the operating pH and which impurities need to be reduced. Biomolecules vary in net charge depending on their isoelectric point (pI). Below the pI, a protein carries a positive charge and binds to a cation exchanger. Above the pI, it carries a negative charge and binds to an anion exchanger. Additional factors such as surface chemistry, particle size, pore size, as well as process requirement also influence resin selection.
POROS ion exchange chromatography resins are supplied in formats intended for use in cGMP production environments and have been implemented in multiple regulatory-approved manufacturing processes. Documentation is available to support regulated processes.
POROS resins use a poly(styrene-divinylbenzene) backbone, which enables reproducible scale-up and chemical stability for cleaning. Their pore structure facilitates high capacity and resolution across a wide range of flow rates. A 50-micron bead size improves resolution with tighter peaks and smaller elution volumes, supporting efficient downstream processing.
Thermo Fisher Scientific offers a range of chromatography products beyond cation exchange resins. Our portfolio includes affinity, anion-exchange, hydrophobic interaction, and mixed-mode resins. These resins can be applied to typical therapeutic biologic workflows such as antibody, protein, nucleic acid, and viral vector processing.
IEX resins are applied across multiple modalities to enable efficient separation and purification based on molecular charge:
They are also used in related applications, such as plasma fractionation and exosome purification. This flexibility helps streamline workflows from process development through cGMP manufacturing.
Yes, samples for evaluation can be requested through the Biotherapeutics Purification Sample Request Form.
Chromatography resources
Access application notes, protocols, and videos that support resin evaluation and process development. Topics include resin selection, column operation, method transfer, and troubleshooting for antibodies, viral vectors, and mRNA. These materials offer guidance for downstream workflows, spanning from early research to manufacturing.
Start your purification journey
Use of the products may vary. For specific use statements, please see product literature.