Eliminate waste and streamline your labeling process by printing cassettes on demand with the Thermo Scientific™ PrintMate™ AS Cassette Printer at the grossing station.
- Save time—print each cassette in ten seconds
- Compact footprint allows the printer to fit easily next to the grossing station
- Print only the cassettes you need, right at the point of use
- Print barcodes on cassettes to automate downstream processes
- Eliminate error-filled handwriting processes
Maximize your efficiency. Print 2-D data matrix barcodes on cassettes to automate sample identification in downstream processes. Barcodes printed on cassettes can contain complete case identification information. Barcodes can be scanned at the microtome to automate slide printing.
Related products
Enhance the Thermo Scientific™ PrintMate™ AS Cassette Printer with these Thermo Scientific™ PrintMate™ AS Cassette Printer Accessories.
Eliminate waste and streamline your labeling process by printing cassettes on-demand at the grossing station with the Thermo Scientific™ PrintMate™ AS Cassette Printer
Everything you need for your SlideMate AS Slide Printer, SlideMate Slide Printer, PrintMate AS Cassette Printer and PrintMate Cassette Printer. Collection trays, marking pens and more.