Slide Basket Transfer System
Save time and steps in your staining and coverslipping by connecting the Gemini AS and ClearVue together with the Slide Basket Transfer System.
In your workflow, saving time is essential. The Thermo Scientific Slide Basket Transfer System seamlessly transfers the slide basket from the Gemini to the ClearVue for you so you can focus on other things.
Stop interrupting your workflow to transfer slide baskets. Simply place your slides in the Gemini AS slide stainer and the Slide Basket Transfer System will automatically transfer them to be coverslipped in the ClearVue coverslipper.
  • Free up your team’s time with automatic transfer of slide baskets from the Gemini AS slide stainer to the ClearVue coverslipper.
  • Simply place the slides in the Gemini AS slide stainer and carry-on with other tasks. Be alerted when the slides have finished coverslipping in the ClearVue coverslipper.  
  • With a small footprint, the Slide Basket Transfer System fits into virtually any laboratory’s workflow.
  • By eliminating transfer between the Gemini AS slide stainer and the ClearVue coverslipper, reduce your team’s exposure to harmful fumes.
  • Compatible with Gemini AS slide stainers and ClearVue coverslippers made after February 2018.