Designed to fit on a benchtop, the compact CTS Rotea system’s counterflow centrifugation technology is designed to gently and efficiently separate cells, aggregates, or beads based on size and to concentrate particles with outer diameter (OD) >5 μm.
The proprietary Draw Extraction Technology lets users gently and precisely recover washed and concentrated cells from the processing chamber. In addition, the CTS Rotea system incorporates multiple sensors to monitor and protect the integrity of the closed system.
Attribute | Description |
Centrifugal force | 3000 x g |
Peristaltic pump flow rate | 5–160 mL/min* |
Centrifuge chamber volume | 10 mL |
Cell count per chamber load | 5 x 10⁷–5 x 10⁹ cells** |
Output cell concentration | 5 x 10⁶–500 x 10⁶ cells/mL |
Recommended input volume range | 50 mL–20 L |
Minimum output volume range | 5 mL |
Maximum fluid density | 1.10 g/mL |
Operating noise level | <70 dBA |
Dimensions (H x W x D) | 16 x 11.4 x 20 in (body only), 30 x 11.4 x 20 in (with bag poles) (40 x 29 x 50.8 cm (body only), 76.2 x 29 x 50.8 cm (with bag poles)) |
Weight | 44 lb (20 kg) |
Tablet accessory model | 3310 Dell system (laptop) |
*Dependent on single-use kit type.
**Dependent on cell type. Centrifuge can be filled multiple times to process larger batches.
Note: Specifications and parameters above are cell type and protocol dependent. Instrument and Consumable Regulatory Support File available upon request at thermofisher.com/regulatory.
Catalog number | Description | Item Count | Price |
---|---|---|---|
A50760* A47679** |
CTS Rotea System + 2-year service | 1 each |
Get a quote |
A50757* A47695** |
CTS Rotea System + 2-year service (including OQ after PM) + IQOQ | 1 each | Get a quote |
* North America, Europe
** Rest of World
The CTS Rotea single-use kit provides flexibility for process development and seamlessly transfer to clinical-scale manufacturing. Closed, single-use kits enable multi batch processing in a shared, grade C clean room leading to cost-effective transfer and scale-out.
Parameter | Specification |
Kit sterility | Compatible with Luer connectors and sterile welding |
Sterilization method | Gamma sterilized |
Tubing material | DEHP-free PVC |
Flow rates | Standard kit: 5–110 mL/min Hi-flow kit: 30–160 mL/min |
Output cell concentration | Up to 500 x 106 cells/mL |
For Research Use or Manufacturing of Cell, Gene, or Tissue-Based Products.