The Xenon Electroporation System is designed for superior non-viral transfection performance, ease of use, and a closed, integrated workflow. Two compatible electroporation configurations allow seamless scaling from process development to commercial manufacturing.
The Xenon electroporation instrument is designed to fit into a closed, integrated cell therapy manufacturing workflow. The ability to adjust electroporation parameters and scale from small to large volume enables efficient process development.
Attribute |
Description |
---|---|
Recommended process volume cell concentration |
20 x 106–100 x 106 cells/mL |
Electroporation volumes |
1 mL; 5–25 mL |
Electroporation chamber volume |
1 mL |
Run time for 5–25 mL electroporation volume |
7–22 min |
Electroporation pulse voltage range |
500–2,500 V |
Electroporation pulse width range |
1–30 ms |
Electroporation pulse interval range |
500–1,000 ms |
Number of electroporation pulses |
1–10 |
Supports 21 CFR part 11 compliance |
Yes, with software upgrade (please inquire) |
Open Platform Communications–Unified Architecture (OPC-UA) compatible |
Yes |
Cloud connect utility |
Yes |
Cell mixer rotation speed |
60 rpm |
Precooling technology |
Peltier |
Precooling temperature setting range |
10–30°C |
Dimensions (H x W x D), door fully opened |
43.1 x 26.5 x 21.2 in |
Weight |
154.3 lb (70 kg) |
Electrical rating |
100–240 V, 1,200 VA |
Touch display |
8 inch capacitive |
Note: Instrument and Consumable Regulatory Support File available upon request at thermofisher.com/regulatory.
To view specifications for consumables, such as the SingleShot electroporation chamber and the MultiShot electroporation cartridge, see Accessories.
Intended use of the products mentioned on this page varies. For specific intended use statements, please refer to the product label.