Coronavirus
Responsive, flexible, and sensitive portfolio of products to fit every need
Thermo Fisher Scientific is an industry-leading, worldwide, trusted partner with comprehensive, end-to-end research solutions for pathogen detection; scientific expertise; and the support capabilities critical to combating the current SARS-CoV-2 outbreak. We are committed to helping you rapidly address the outbreak with ready-to-use qPCR workflow research products, or the individual research components, referenced in CDC and WHO protocols.
For Research Use Only. Not for use in diagnostic procedures.
For the most up-to-date information on the current coronavirus outbreak, visit the WHO or CDC websites.
Now available: Singplex and multiplex assay kits for SARS-CoV-2 pathogen detection (For Research Use Only)
Our newly developed real-time PCR kits for SARS-CoV-2 pathogen detection research are now available to order. Both singleplex and multiplex kit workflows were developed for use on both the Applied Biosystems QuantStudio 5 and 7500 real-time PCR systems. The kit utilizes Applied Biosystems TaqMan Assay technology, enabling specific, sensitive, and accurate results.
TaqMan 2019-nCoV Assay Kit v1 (singleplex)* TaqMan SARS-CoV-2 Assay Kit v2 (multiplex)
  • Singlexplex CoV Assay, for customers with low throughput samples
  • One coronavirus gene per well
  • FAM Dye
  • Kit includes four wet tubes containing assays, delivering 50 reactions
  • Control kit also available
  • Utilizes TaqPath 1-Step RT-qPCR Master Mix, CG 
  • Multiplex CoV Assay, for customers with high throughput samples
  • Three coronavirus genes per well
  • Multiple dyes
  • Available in both 100 and 1,000 reaction formats
  • SARS-CoV-2 DNA and MS2 bacteriophage control kits available
  • Utilizes TaqPath 1-Step Multiplex Master Mix (No ROX)
Order now › Order now ›
* Although the singleplex kit has “2019-nCoV” in its product name, both kits detect the SARS-CoV-2 pathogen. The kit was named and catalogued prior to recent WHO naming updates.
Both kits feature assays specifically targeted to all >100 complete genomes currently available at GISAID, and do not target any of the 2,116 complete genomes of other coronaviruses currently available at NCBI.
Product Components Quantity Cat. no. Documents
Singleplex solution
TaqMan 2019-nCoV Assay Kit v1 2019-nCoV Assay (orf1ab); 2019-nCoV Assay (spike (S) gene); 2019-nCoV Assay (nucleocapsid (N) gene); Rnase P (control) 50 reactions A47532 Protocol
TaqMan 2019-nCoV Control Kit v1   2019-nCoV Control (covering orf1ab, spike (S) gene, nucleocapsid (N) gene, RNaseP) 50 µL (1 x 10copies/µL) A47533 Protocol
Multiplex solution
TaqMan SARS-CoV-2 Assay Kit v2, 100 Reactions SARS-CoV-2 Assay Multiplex (Gene Orf-1ab, N Protein, S Protein, MS2) 100 reactions (150 μL) Get a quote
TaqMan SARS-CoV-2 Assay Kit v2, 1000 Reactions SARS-CoV-2 Assay Multiplex (Gene Orf-1ab, N Protein, S Protein, MS2) 1,000 reactions (1500 μL) Get a quote
TaqMan SARS-CoV-2 Control Kit v2 SARS-CoV-2 DNA Control (covering Gene Orf-1ab, Gene S protein and Gene N protein) 250 μL (1 x 104 copies/μL) Get a quote
TaqMan MS2 Phage Control MS2 Phage Control 250 μL (1 x 106 conc) Get a quote
‡ Protocol available upon request for quote.
End-to-end singleplex and multiplex research solutions for SARS-CoV-2 pathogen detection by real-time PCR*
Recommended qPCR master mixes for use with singleplex and multiplex kits for SARS-CoV-2 pathogen detection
Product
Quantity
Cat. no.
Use with:
TaqPath 1-Step RT-qPCR Master Mix, CG  (for use with TaqMan 2019-nCoV Assay Kit v1) 5 x 1 mL A15299 TaqMan 2019-nCoV Assay Kit v1 (singleplex)
10 mL A15300
TaqPath 1-Step Multiplex Master Mix (No ROX) 1 x 0.5 mL A28521 TaqMan SARS-CoV-2 Assay Kit v2 (multiplex)
1 x 10 mL A28523
5 x 1 mL A28522
Recommended qPCR systems for use with singleplex and multiplex kits for SARS-CoV-2 pathogen detection research
7500 Fast and 7500 real-time PCR systems
Applied Biosystems 7500 real-time PCR systems offer high performance, multicolor real-time PCR wherever your research takes you. Choose from our Fast (0.1 mL) and standard (0.2 mL) 7500 systems. All Applied Biosystems 7500 systems are designed to deliver the reliable performance and dye flexibility you need to generate data that you can trust.
QuantStudio 5 Real-Time PCR System (0.1- and 0.2-mL 96-well blocks, 384-well block)
The Applied Biosystems QuantStudio 5 Real-Time PCR System is ideal for all levels of users who need a high-performance instrument with features for maximum control of their experiments. Includes built-in security features, locked workflows, user account management, and options for web browser–based or desktop analysis.
Learn more about our real-time PCR systems ›
Thermo Fisher Scientific products referenced in publicly available real-time PCR protocols for SARS-CoV-2 pathogen detection
Related products
Real-time PCR and 1-step RT-PCR master mixes
Product Description Cat. no.
TaqMan Fast Virus 1-Step Master Mix
  • Highly concentrated (4X) master mix—add more sample for improved sensitivity
  • Increased tolerance to inhibitors for greater accuracy
  • One-tube master mix for improved efficiency and ease of use
  • Optimized for multiplexing (up to 3 targets) and with exogenous/endogenous internal controls
  • Fast cycling—provides results in ~60 min, half the time of standard cycling (also works with standard cycling)
4444434
TaqPath 1-Step RT-qPCR Master Mix, CG All the advantages of TaqMan Fast Virus 1-Step Master Mix plus:
  • Includes UNG and dUTP for carryover contamination control
  • Enhanced quality control measures for even better lot-to-lot consistency
  • General purpose reagent
A15299  
SuperScript III Platinum One-Step qRT-PCR Kit
  • Delivers reliable, accurate target detection
  • Uses SuperScript III reverse transcriptase (RT) for higher temperature cDNA synthesis to overcome RNA secondary structure
  • Features Platinum Taq DNA Polymerase with hot-start technology for improved specificity
  • Verified performance on multiple real-time instrument platforms
11732020
SuperScript III One-Step RT-PCR system with Platinum Taq DNA Polymerase
  • Sensitive—routine detection down to 0.01 pg total RNA
  • Convenience—one-step format for speed, convenience, and less reaction-to-reaction variability
  • Specificity—uses SuperScript III RT for cDNA synthesis up to 55°C, for more specific priming with gene-specific primers
  • Amplicon size—compatible detection of targets up to 4.5 kb in length for greater flexibility
12574018
Viral nucleic acid isolation products
Isolate RNA from SARS-CoV-2 quickly and efficiently using the Applied Biosystems MagMAX Viral/Pathogen Nucleic Acid Isolation Kit and Thermo Scientific KingFisher purification instruments.
Isolate high-quality DNA and RNA with the MagMAX Viral/Pathogen Nucleic Acid Isolation Kit. Features of this kit include:
  • Automation-ready protocols designed for the KingFisher Flex and KingFisher Duo Prime systems
  • Fast procedure allows for 96 samples to be processed in <30 minutes
  • Flexible protocol accommodates research sample volume inputs from 200 µL to 2 mL of blood, urine, swabs, lavage, and various transport media such as universal viral transport medium, and Eswab preservative fluid
  • Compatible with bronchoaveolar lavage and other respiratory research samples
  • Elution volumes ranging from 50 to 100 µL
Automate your DNA and RNA isolation with the KingFisher Flex system, a versatile benchtop instrument that processes 24 or 96 samples per run. Features of this system include:
  • High-throughput purification of 24 or 96 samples per run
  • Easy to install and run (ready to run in 10 minutes or less)
  • Choose from two plate formats for wide volume range (20–5,000 µL)
  • Download existing protocols from library, or easily customize protocols using Thermo Scientific BindIt Software
For Research Use Only. Not for use in diagnostic procedures.