EGFR IHC kit features
- Complete and standardized kit for immunohistochemistry
- For IHC staining of formalin-fixed, paraffin-embedded tissues
- For identification of EGFr protein expression in normal and neoplastic tissues
- SuperPicture™ polymer technology: faster, cleaner and easier than a LAB-SA method
3+ EGFr expression in colon cancer.
3+ EGFr expression in colon cancer.
EGFR IHC kit description
EGFr IHC kit features the EGFr antibody clone 31G7, which is an industry standard for the detection of EGFr in formalin-fixed, paraffin-embedded tissue samples.1-5 The highly specific clone 31G7 recognizes the extracellular domain of EGFr, and does not cross react with the related erbB receptor family. 31G7 has been widely used to determine the immunoreactive EGFr present in colon cancers and squamous cell carcinomas that arise from both squamous epithelium & metaplastic squamous epithelium. EGFr IHC kit uses proprietary SuperPicture™ polymer technology, which is highly sensitive, and yields intense staining without background. The EGFr kit is a complete IHC kit, including the prediluted EGFr antibody, pretreatment reagent, polymer detection system, Mouse IgG negative isotype control, 5 control slides, and a color evaluation guide.
References
- Fagbemi AO, et al. Early Human Dev 65: 1-9, 2001.
- Leav I, et al. Human Pathol 29: 668-75, 1998.
- Niikura H, et al. Human Pathol 27: 282-9, 1996.
- Elias JM, et al. J Histotechnology 19(1): 13-22, 1996.
- Coffin CM, et al. Arch Pathol Lab Med 120: 1032-8, 1996.
- Reyes A, et al. Am J Clin Pathol 120:182-187, 2003.
- Rusch VW, et al. Ann Thorac Surg 62(3):798-810, 1996.
- Cappuzzo F, et al. J Clin Oncol 21:2658-2663, 2003.
- Choe G, et al. Cancer Res 63:2742-2746, 2003.
- Liu TF, et al. Cancer Res 63:1834-1837, 2003.
- Ding H, et al. Cancer Res 63:1106-1113, 2003.
- Costa MJ, and Walls J. Cancer 77(3):533-542, 1996.
- Piyathilake CJ, et al. Clin Cancer Res 8(3):734-744, 2002.
- Parker C, et al. J Histochem Cytochem 46(5):595-602, 1998.
Find Antibodies
Search by your protein of interest, then filter by validated application, reactivity, conjugate, or host to find exactly the antibody you need.