Now with Superior Performance and Increased Stability
New, next-generation detection products for immunohistochemistry (IHC) feature improved DAB substrate reagents for greater signal sensitivity and the addition of an endogenous peroxidase quenching solution for a more complete IHC staining protocol. Increased sensitivity allows clear target antigen detection and higher primary antibody dilution factors.
  • Provide more sensitivity and more consistent staining with difficult-to-detect antigens
  • Offer greater stability and ease of use
  • Save money by increasing primary antibody dilutions
Increased Sensitivity
Mouse anti-EGFr Stained
Figure 1. Mouse anti-EGFr stained esophageal carcinoma tissue sections.
(A) Image using the improved SuperPicture 3rd Gen IHC Detection Kit.
(B) Image using a leading competitor’s kit. Note the intense staining and distinct labeling of the cellular membrane in Image A.
Increased Stability
Mouse anti-PCNA Stained
Figure 2. Mouse anti-PCNA stained small intestine tissue sections.
(A) Image using the new 2-solution DAB that was freshly prepared prior to applying to the section.
(B) Image using the 2-solution DAB after storage for 30 days at 37ºC, and then mixed for use. Note the clear nuclear staining and similar IHC performance without any increase in background staining.
Increased User Tolerance and Ease of Use
Mouse anti-PCNA Stained Tonsil Tissue
Figure 3. Mouse anti-PCNA stained tonsil tissue section. The 2-solution DAB was freshly prepared prior to applying to the section.
Image (A) was taken after incubating for 5 minutes,
Image (B) after incubating for 20 minutes, and
Image (C) after incubating for 40 minutes.
The new DAB solution is more user tolerant, and much less likely to result in high background staining.
Ordering Information
Product Cat. no. Kit contents
SuperPicture™ 3rd Gen IHC Kit (150 slides) 879673*
  • 16.5 ml Peroxidase quenching solution
  • 16.5 ml HRP polymer conjugate
  • 30 ml DAB substrate buffer
  • 1 ml DAB chromogen
SuperPicture™ 3rd Gen IHC Kit (1,000 slides) 878973*
  • 110 ml Peroxidase quenching solution
  • 110 ml HRP polymer conjugate
  • 110 ml DAB substrate buffer
  • 3.5 ml DAB chromogen
Histostain®-Plus 3rd Gen IHC Kit (150 slides) 859673*
  • 16.5 ml Peroxidase quenching solution
  • 16.5 ml Blocking solution
  • 16.5 ml Biotinylated secondary antibody
  • 16.5 ml Streptavidin-peroxidase conjugate
  • 30 ml DAB substrate buffer
  • 1 ml DAB chromogen
Histostain®-Plus 3rd Gen IHC Kit (1,000 slides) 859073*
  • 110 ml Peroxidase quenching solution
  • 110 ml Blocking solution
  • 110 ml Biotinylated secondary antibody
  • 110 ml Streptavidin-peroxidase conjugate
  • 110 ml DAB substrate buffer
  • 3.5 ml DAB chromogen
2-Solution DAB Kit (1,000 slides) 882014*
  • 110 ml DAB substrate buffer
  • 3.5 ml DAB chromogen
* For in vitro diagnostic use