Gibco® reagents for neural cell culture have been trusted and published by researchers for more than twenty years.  This trusted family of products is available  under the CTS™ brand to provide workflow solutions that will:
  • Reduce your burden in qualifying reagents during transition from research to clinical applications
  • Easily meet regulatory and quality requirements
Learn more about what CTS™ products offer
We offer a collection of Cell Therapy Systems (CTS™) products for cell therapy research and further manufacturing of cell-, gene- and tissue-based products. For more information about our product offerings, ordering information and customer support, access our Gibco® CTS™ brochure: View brochure.
Request more information  
Neurobasal® CTS™ Media
Ideal for maintenance and expansion of human embryonic or post-natal neural stem cells and primary neurons. Allows for pure populations of neuronal cells without the need for an astrocyte feeder layer.
View Product Details
B-27® Supplement, XenoFree CTS™
Xenofree variation of the original, trusted B-27® Serum Free Supplement.  All animal origin components from the original formulation have been replaced with recombinant or human origin components.
Ideal for studies requiring the absence of non-human origin components, such as differentiation or neural stem cells.
View Product Details
N-2 Supplement CTS™
Serum-free, chemically defined supplement for the growth and expression of post-mitotic neurons and tumor cells of neuronal phenotype. 
  • Create a complete media by using N-2 Supplement CTS™ with any Neurobasal® CTS™ or KnockOut™ DMEM/F-12 CTS™  media.
View Product Details
Hibernate® CTS™ Media
Serum-free, nutrient basal media for short-term maintenance of CNS tissues and cells at ambient CO2 conditions.
  • Use Hibernate®-A CTS™ Medium with postnatal neurons
  • Use Hibernate®-E CTS™ Medium with embryonic neurons
View Product Details
Synth-a-Freeze® CTS™ media
Chemically defined, protein-free liquid cryopreservation medium intended for freezing and storing a variety of mammalian cell types including neural stem cells.
View Product Details
Gibco® CTS™ Growth Factors
High quality growth factors and cytokines for t-cell and dendritic cell applications.
View Product Details
CELLstart™ CTS™ substrate
Fully defined, xenofree substrate for neural stem cell growth and expansion.  Dilute with DPBS CTS™ media with calcium and magnesium.
View Product Details
For Research Use or Non-Commercial Manufacturing of Cell- Based Products for Clinical Research. CAUTION: Not Intended for Direct Administration into Humans or Animals.
Neural Cell Therapy Resources
 CTS Product Support
Cell therapy support