Order online
Order QuantiGene Plex DNA CNV assays using the tool below. Search our catalog of thousands of probe sets below, and if your gene of interest is not found, we can design a new probe set within 5-7 days. Contact your sales specialist or email genomicorders@thermofisher.com or customercare@thermofisher.com for more information.
Find your target species and your desired gene of interest
QuantiGene Plex Assay Kits contain the buffers, plates, detection reagents to perform the assay.
Description | Product size | Cat. No. |
---|---|---|
QuantiGene Plex DNA Assay Kit | 1 plate | QPD1010 |
QuantiGene Plex DNA Assay Kit | 3 plate | QPD1011 |
QuantiGene Plex DNA Assay Kit | 10 plate | QPD1012 |
By request QuantiGene Plex DNA panels
"By Request" Plex Sets are also available (3 to 80-plex) and in 1, 3 or 10 plate quantities. Catalog plexes can also be modified. We can design new targets/panels for a $185 development fee per new target. If the target already exists in our catalog, there are no additional fees. You can contact your local sales manager for a quote.
Catalog QuantiGene Plex Sets contain two components, a target-specific Probe Set and Magnetic Capture Beads, which are premixed and ready to use with a QuantiGene Plex DNA Assay Kit. All Plex Sets are available in 3-sizes, 1-, 3- and 10-plate. Select from any of our existing panels or any subset of targets from that panel, your panel will be delivered within 5 working days.
QuantiGene Sample Processing Kits are designed for a variety of sample types and contain the reagents and instructions for the preparation of DNA from cultured cells, tissues, FFPE tissues, and blood. These kits are not required if working with purified DNA.
Description | Product size | Cat. No. |
---|---|---|
QuantiGene Sample Processing Kit for Cultured Cells | 2 plates | QS0100 |
QuantiGene Sample Processing Kit for Cultured Cells | 10 plates | QS0101 |
QuantiGene Sample Processing Kit for Cultured Cells | 5 x 10 plates | QS0102 |
QuantiGene Sample Processing Kit for Cultured Cells | 50 plates | QS0103 |
QuantiGene Sample Processing Kit for Tissues | 10 samples | QS0104 |
QuantiGene Sample Processing Kit for Tissues | 25 samples | QS0105 |
QuantiGene Sample Processing Kit for Tissues | 100 sample | QS0106 |
QuantiGene Sample Processing Kit for FFPE Samples | 10 samples | QS0107 |
QuantiGene Sample Processing Kit for FFPE Samples | 25 samples | QS0108 |
QuantiGene Sample Processing Kit for FFPE Samples | 100 samples | QS0109 |
QuantiGene Sample Processing Kit for Blood Samples | 2 plates | QS0110 |
QuantiGene Sample Processing Kit for Blood Samples | 10 plates | QS0111 |
QuantiGene Sample Processing Kit for Blood Samples | 5 x 10 plates | QS0112 |
Genomic DNAs
Recommended heated, plate shaking incubator for the QuantiGene Plex assay. Provides optimal temperature uniformity during the hybridization steps
Description | Product size | Cat. No. |
---|---|---|
Control Genomic DNA, Human Male | 10 ug | QPD0600 |
Control Genomic DNA, Human Female | 10 ug | QPD0601 |
Control Genomic DNA, Rat Male | 10 ug | QPD0602 |
Temperature validation kit
The QuantiGene Incubator Temperature Validation Kit uses a calibrated thermometer to assess the accuracy of the temperature of the oven incubator used in the assay.
Description | Product Size | Cat. No. |
---|---|---|
QuantiGene Temperature Validation Kit | 1 kit | QS0517 |
Hand-held magnetic plate washer
The Hand-Held Magnetic Plate Washer is designed for use with the QuantiGene Plex assay configured for magnetic separation.
Description | Product size | Cat. No. |
---|---|---|
Hand Held Magnetic Plate Washer- QGP | each | QP0702 |
VorTemp plate shaking incubator
Recommended heated, plate shaking incubator for the QuantiGene Plex assay. Provides optimal temperature uniformity during the hybridization steps
Description | Product size | Cat. No. |
---|---|---|
VorTemp 56, 120V- holds 2 plates | each | QP0703 |
VorTemp 56, 220V- holds 2 plates | each | QP0704 |
QuantiGene Plex supplemental components
Description | Product size | Cat. No. |
---|---|---|
QGP Hybridization Plate | 10 each | QP0545 |
QGP Magnetic Separation Plate | 10 each | QP0540 |
QGP Pressure Seals (use with hybridization plate) | 12 each | QP0544 |
QuantiGene Foil Plate Seals (use with Mag Sep Plate) | 12 each | QS0506 |
QuantiGene Foil Plate Seals (use with Mag Sep Plate | 260 each | QS0507 |
QuantiGene Foil Plate Seals | 45 each | QG0524 |
QuantiGene Homogenizing Solution | 20 mL | QS0518 |
QuantiGene Homogenizing Solution | 50 mL | QG0516 |
QuantiGene Homogenizing Solution | 200 mL | QG0517 |
QuantiGene Plex Lysis Mixture | 12 mL | QP0522 |
QuantiGene Plex Lysis Mixture | 36 mL | QP0523 |
QuantiGene Plex Lysis Mixture | 120 mL | QP0524 |
Proteinase K, 50 µg/µL | 150 µL | QS0510 |
Proteinase K, 50 µg/µL | 750 µL | QS0511 |
Proteinase K, 50 µg/µL | 3.5 mL | QS0512 |
QuantiGene Plex Wash Buffer Components | 1 plate | QP1501 |
QuantiGene Plex Wash Buffer Components | 3 plates | QP1502 |
QuantiGene Plex Wash Buffer Components | 10 plates | QP1503 |
For Research Use Only. Not for use in diagnostic procedures.