Over seven million samples analyzed to date—and growing
Combining the power of the QuantStudio 12K Flex Real-time PCR System with qualified TaqMan genotyping assays, this solution delivers a sample-to-answer workflow that allows you to introduce pharmacogenomic testing into your laboratory with ease.
Features include:
- Fully qualified content—all TaqMan SNP Genotyping Assays have been tested against genomic DNA and synthetic controls
- Express Panels include our most popular PGx content including—CYP1A2, CYP2B6, CYP2C9, CYP2C19, CYP2D6, CYP2E1, CYP3A4, CYP3A5, SLCO1B1, VKORC1, COMT, OPRM1, DRD2, FII, FV, MTHFR, APOE2, APOE4
- Flexible formats—our PGx solution has been optimized for anything from custom content to fit your needs, to off-the shelf content
- Extensive menu of targets—over 2,700 TaqMan SNP Genotyping Assays available, targeting the most common drug metabolizing enzyme (DME) markers (including CYP2D6, CYP2C9, CYP4F2, and more)
- Calling and reporting solutions—TaqMan Genotyper Software and AlleleTyper Software are available* to quickly and easily determine haplotypes of samples using star-allele nomenclature
- Utilizing familiar instruments—our PGx solutions can be run on either QuantStudio 5 or QuantStudio 7 for custom plates, and QuantStudio 12K Flex System for OpenArray
*Additional PGx data and support files at bottom of page
TaqMan OpenArray (OA) | TaqMan Array Plates (TAP) | |
---|---|---|
Focus | For mid-higher throughput applications | For low-mid throughput applications |
Targets | TaqMan OpenArray PGx Express – 60:
| 32 assays x 12 samples: 80 plates 64 assays x 6 samples: 160 plates 128 assays x 3 samples: 320 plates |
Goals/needs |
|
|
Format | Off-the-Shelf and Custom 2,880 reactions with pre-spotted assays 4-hour workflow | Off-the-Shelf and Custom 384-well plates with pre-spotted assays PGx Express (60) on 384-well plates 3-hour workflow |
Compatible instruments | QuantStudio 12K Flex Real-Time PCR System with OpenArray block | QuantStudio 12K Flex with 384-well block QuantStudio 7 Flex & 7 Pro with 384-well block QuantStudio 6 Flex & 6 Pro with 384-well block QuantStudio 5 with 384-well block |
Product | Unit Size | Catalog Number | Ordering information |
1 kit | 4404846 | ||
Sizes vary | Multiple SKUs | ||
Sizes vary | Multiple SKUs |
We offer a simple, efficient workflow designed to maximize results.
Buccal swab, saliva, or blood
Thermo Scientific™ KingFisher™ Flex system with Applied Biosystems™ MagMAX™ DNA Multi-Sample Ultra Kit
Applied Biosystems™ OpenArray™ AccuFill™ System
Applied Biosystems™ QuantStudio™ 12K Flex Real-Time PCR System
Drug metabolism enzyme (DME) assays run on OpenArray plates or 384-well plates
Genotyping data: In-house laboratory information systems (LIS) or relative quantification application
Buccal swab, saliva, or blood
Thermo Scientific™ KingFisher™ Flex system with Applied Biosystems™ MagMAX™ DNA Multi-Sample Ultra Kit
Applied Biosystems™ OpenArray™ AccuFill™ System
Applied Biosystems™ QuantStudio™ 12K Flex Real-Time PCR System
Drug metabolism enzyme (DME) assays run on OpenArray plates or 384-well plates
Genotyping data: In-house laboratory information systems (LIS) or relative quantification application
For Research Use Only. Not for use in diagnostic procedures.