Search
Immunosuppressant drug monitoring (ISD) assays are crucial for monitoring immunosuppressant drug levels in transplant patients to prevent organ rejection. We offer a full line of ISD assays, including cyclosporine, mycophenolic acid, tacrolimus, and our exclusive everolimus, to aid in patient monitoring.
These immunoassays, referenced to LC-MS/MS or HPLC/MS—the gold standards—are precise, sensitive, and specific. Applications are available on our Thermo Scientific Indiko™ and Indiko™ Plus clinical and specialty chemistry analyzers, complete with convenient bar-coded reagents as well as a wide variety of other laboratory instrumentation.
Achieve precise and accurate quantitative results crucial for monitoring drug levels of cyclosporine in kidney, liver and heart transplant patients with our easy-to-use Thermo Scientific™ CEDIA™ Cyclosporine Plus Immunoassay.
Complete your immunosuppressant drug testing panel with the Thermo Scientific™ CEDIA™ Mycophenolic Acid Immunoassay. Like our other ISD assays, the CEDIA Mycophenolic Acid Immunoassay is accurate and precise, providing users with confidence in their results. Applications are available on our flexible and intuitive Indiko™ and Indiko Plus clinical and specialty chemical systems as well as a wide variety of other laboratory instrumentation.
Obtain results you can always count on for the crucial monitoring of everolimus drug levels in kidney and liver transplant patients with the Thermo Scientific™ QMS™ Everolimus Immunoassay. Using our proprietary QMS technology, these liquid, ready-to-use reagents provide excellent accuracy and precision. Applications are available on our compact Indiko™ and Indiko Plus clinical and specialty chemistry systems as well as a wide variety of other laboratory instrumentation.
Achieve high sensitivity and excellent low-end precision when monitoring kidney, liver and heart transplant patients with the Thermo Scientific™ QMS™ Tacrolimus Immunoassay. With a reportable range down to 1ng/mL, our Tacrolimus Immunoassay is the perfect aid for monitoring patients on multiple drug therapies. The reagents are liquid and ready-to-use with applications available for our fast and efficient Indiko™ and Indiko Plus clinical and specialty chemistry systems or on a wide variety of other laboratory instrumentation.
Methodist Dallas Medical Center performs hundreds of liver and kidney transplants per year. With such volume, the medical center must be hyper-vigilant about lab result quality and efficiency.