One Lambda MagSort isolates specific HLA Antibodies using magnetic bead panels to adsorb and elute targeted antibodies.
One Lambda™ MagSort™ addresses emerging interests and challenges in adsorption and elution techniques for HLA antibody testing. Targeting individual HLA antigens enables a more focused dataset, to explore and define complex HLA antibody profiles at the antigen-specific level. This novel technology streamlines the process, reducing hands-on time for the laboratory staff, and standardizes a unique isolation approach compared to more cumbersome cell-based approaches.
Capabilities
Targeted technology
  • Individual antigen-coated magnetic beads
  • Elute antibodies that bind to a specific antigen
  • Results in a more focused HLA antibody assessment 
Customizable panels  
  • 59 antigens across 6 locus specific kits
  • Flexibility to focus only on antigens of interest
  • Coverage across well-documented antigens
Efficient workflow 
  • Up to 45 minutes of hands-on time and as little as 4 hours to      workflow completion
  • Uses equipment already found in most laboratories
  • Immediate antigen availability.
How does it work?
MagSort Workflow
MagSort example of results before and after isolating A*02:01
Products & coverage
MagSort Class I HLA-A
MagSort Class I HLA-B
MagSort Class I HLA-C
MagSort Class II HLA-DRB1  MagSort Class II HLA-DQ MagSort Class II HLA-DP
A*01:01
B*07:02
C*01:02
DRB1*01:01 DQB1*02:01
DQA1*02:01
DPB1*01:01
DPA1*01:03
A*02:01
B*08:01
C*02:02
DRB1*03:01
DQB1*02:02
DQA1*02:01
DPB1*02:01
DPA1*01:03
A*03:01
B*13:01
C*03:03
DRB1*04:01
DQB1*03:01
DQA1*02:01
DPB1*03:01
DPA1*01:03
A*11:01
B*15:01
C*03:04
DRB1*04:04
DQB1*03:02
DQA1*02:01
DPB1*04:01
DPA1*01:03
A*24:02
B*44:02
C*04:01
DRB1*07:01
DQB1*03:03
DQA1*02:01
DPB1*04;02
DPA1*01:03
A*25:01
B*46:01
C*05:01
DRB1*08:01
DQB1*04:01
DQA1*02:01
DPB1*11:01
DPA1*01:03
A*29:01 B*53:01 C*07:02 DRB1*11:01 DQB1*05:01
DQA1*01:01
DPB1*17:01
DPA1*02:01
A*30:01 B*57:01 C*12:03 DRB1*12:01 DQB1*06:02
DQA1*01:01
DPB1*19:01
DPA1*01:03
A*33:03 B*67:01 C*15:02 DRB1*13:01 DQB1*06:03
DQA1*01:03
A*68:01 B*73:01 C*16:01 DRB1*15:01    
    C*17:01 DRB1*16:01    
Need help finding the right solution?