Informational:As of April 26, 2023 Accula products are no longer available for direct purchase through Thermo Fisher Scientific. Authorized distributors will continue to sell the Accula products until inventory runs out.
Thermo Fisher Scientific
Accula™ SARS-CoV-2 Test
Catalog number: COV4100
Thermo Fisher Scientific
Accula™ SARS-CoV-2 Test
Catalog number: COV4100
Catalog Number
COV4100
Unit Size
25 pack
Price (USD)
Price: 1,562.50
Catalog NumberUnit SizePrice (USD)
COV410025 pack
Price: 1,562.50
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The Accula SARS-CoV-2 Test combines RT-PCR accuracy with a simple workflow to provide fast and convenient rapid PCR-based test results. The test’s patented PCR technology enables rapid exponential amplification while reducing overall thermocycling times, resulting in faster test results. 1
The Accula SARS-CoV-2 Test performed on the Accula Dock is a molecular in vitro diagnostic test utilizing polymerase chain reaction (PCR) and lateral flow technologies for the qualitative, visual detection of nucleic acid from SARS-CoV-2 in clinician-collected anterior nasal or nasal mid-turbinate swab specimens or clinician-instructed self-collected (collected on site) anterior nasal swab specimens, collected from individuals suspected of COVID-19 by their healthcare provider.
Features of the Accula SARS-CoV-2 Test include:
  • Rapid testing results—RT-PCR SARS-CoV-2 results in ∼30 minutes
  • Streamlined process—CLIA-waived workflow
  • Simplified sample collection—anterior nasal swabs and nasal mid-turbinate swabs
  • Easy—store at room temperature, no need for refrigeration
The Accula SARS-CoV-2 Test is a nucleic acid amplification test (NAAT) for detection of SARS-CoV-2 viral RNA in approximately 30 minutes. To perform the test, anterior nasal or nasal mid-turbinate specimens are added to the SARS-CoV-2 buffer to solubilize the sample. An aliquot of the SARS-CoV-2 buffer is then dispensed into an Accula SARS-CoV-2 test cassette. The test cassette contains internal positive and negative controls, enzymes, OscAR™ reagents, and a detection strip necessary for the full completion of the assay. There are four steps in the process:
1. Lysis of the virus
2. Reverse transcription (RT) of viral RNA to cDNA
3. Nucleic acid amplification by PCR
4. Detection
In approximately 30 minutes, clinicians can quickly and reliably identify patients who are positive for COVID-19. Mutations in the following SARS-CoV-2 Variants have been demonstrated to have no impact on the performance of the Accula SARS-CoV-2 Test:2
  • Alpha: B.1.1.7
  • Beta: B.1.351
  • Delta: B.1.617.2
  • Gamma: P.1 
  • Omicron: B.1.1.520, B.1.1.529, BA.1, BA.1.1, BA.2, BA.2.10, BA.2.12, BA.2.12.1, BA.2.75, BA.2.75.2, BA.3, BA.4, BA.4.6, BA.5, BA.5.2.6, BF.7, BF.11, BN.1, BQ.1, BQ.1.1, XBB, and XBB.1.5  
Intended use
The Accula™ SARS-CoV-2 Test performed on the Accula Dock or the Silaris™ Dock is a molecular in vitro diagnostic test utilizing polymerase chain reaction (PCR) and lateral flow technologies for the qualitative, visual detection of nucleic acid from SARS-CoV-2 in clinician-collected anterior nasal or nasal mid-turbinate swab specimens or clinician-instructed self-collected (collected on site) anterior nasal swab specimens, collected from individuals suspected of COVID-19 by their healthcare provider. Testing is limited to laboratories certified under the Clinical Laboratory Improvement Amendments of 1988 (CLIA), 42 U.S.C. §263a, that meet requirements to perform high, moderate, or waived complexity tests. The Accula SARS-CoV-2 Test is authorized for use at the Point of Care (POC), i.e., in patient care settings operating under a CLIA Certificate of Waiver, Certificate of Compliance, or Certificate of Accreditation. 
Accula SARS-CoV-2 Test results are for the identification of SARS-CoV-2 RNA. The SARS-CoV-2 RNA is generally detectable in upper respiratory specimens during the acute phase of infection. Positive results are indicative of the presence of SARS-CoV-2 RNA; clinical correlation with patient history and other diagnostic information is necessary to determine patient infection status. Positive results do not rule out bacterial infection or co-infection with other viruses. Testing facilities within the United States and its territories are required to report all results to the appropriate public health authorities. 
Negative results should be treated as presumptive and, if inconsistent with clinical signs and symptoms or necessary for patient management, should be confirmed with a different authorized or cleared molecular test in a CLIA-certified laboratory that meets the requirements to perform high or moderate complexity tests. Negative results do not preclude SARS-CoV-2 infection and should not be used as the sole basis for patient management decisions. Negative results must be combined with clinical observations, patient history, and/or epidemiological information. 
The Accula SARS-CoV-2 Test is intended for use by trained operators who are proficient in performing tests on the Accula Dock and Silaris Dock. The Accula SARS-CoV-2 Test is only for use under the Food and Drug Administration’s Emergency Use Authorization.
The Accula SARS-CoV-2 Test Kit has not been FDA cleared or approved but has been authorized for emergency use by the FDA for use by laboratories certified under the Clinical Laboratory Improvement Amendments (CLIA) of 1988, 42 U.S.C. §263a, that meet requirements to perform high, moderate, or waived complexity tests. The test is authorized for use at the Point of Care (POC), i.e., in patient care settings operating under a CLIA Certificate of Waiver, Certificate of Compliance, or Certificate of Accreditation. This test has been authorized only for the detection of nucleic acid from SARS-CoV-2, not for any other viruses or pathogens. The emergency use of this test is only authorized for the duration of the declaration that circumstances exist justifying the authorization of emergency use of in vitro diagnostics for detection and/or diagnosis of COVID-19 under Section 564(b)(1) of the Federal Food, Drug, and Cosmetic Act, 21 U.S.C. § 360bbb-3(b)(1), unless the declaration is terminated or authorization is revoked sooner.
For Emergency Use Authorization (EUA) Only. For prescription use only. For in vitro diagnostic use.
1 Refer to IFU
2 Test reports are available through Thermo Fisher Scientific
Please note that the most recent revision of the Instructions for use (IFU) applicable for all Accula SARS-CoV-2 Test kit is available via the link below.  However, there may be some Accula SARS-CoV-2 Kit in your possession which would have the negative control swab manufactured differently. The difference in manufacturing process has no impact to the performance or safety of the negative control swab.
Specifications
Certifications/Compliance
FDA Emergency Use Authorized (EUA)
Control Sets
High, Low, Negative
CE Marker
No
Clia Complexity
Waived
Detectable Analytes
SARS-CoV-2 RNA
DoA Calibrators
No
Format
Test Cassette Kit
Sample Type
Anterior Nasal swab
Sensitivity
Product LoD is 150 copies/mL with inactivated virus
Type
SARS-CoV-2 Assay
For Use With (Application)
RT-PCR, Rapid PCR
Organism Group
Virus
Quantity
25
Shelf Life
Up to 18 months
Test Time
∽30 min.
Contents & Storage
  • Collection swabs (25): sterile swabs for nasal sample collection
  • SARS-CoV-2 buffer (25): single-use vial of solution containing 5 mL of buffer
  • Transfer pipette (25): single-use, fixed volume pipette used to transfer sample from the SARS-CoV-2 Buffer vial into the test cassette. Extra pipettes provided for your convenience.
  • Accula SARS-CoV-2 test cassette (25): single-use, foil-pouched with desiccant and test cassette containing lyophilized reagents for the targeted amplification and detection of viral nucleic acid
  • SARS-CoV-2 high positive, low positive, and negative control swab (1 each): DNA-based synthetic oligo dried onto a swab well above the limit of detection of the test
Figures
Documents & Downloads
Certificates
    Frequently asked questions (FAQs)
    Citations & References
    Search citations by name, author, journal title or abstract text