Search
In recent years, the field of hematology has witnessed remarkable advancements, particularly in rapid next-generation sequencing (NGS). These innovations have significantly enhanced the way myeloid and lymphoid malignancies samples are evaluated. A recent symposium highlighted these advancements through the work of Robert Baker, who has been at the forefront of implementing rapid NGS in his laboratory. This article delves into the key points of Baker’s presentation at the 2024 Association of Molecular Pathology (AMP) meeting, illustrating how these advancements have improved accuracy and turnaround times for test results.
Robert Baker
Molecular Haematology Operations Manager
Health Services Laboratory (HSL)
Health Services Laboratories (HSL) is United Kingdom based clinical laboratory that provides a comprehensive range of diagnostic services, including pathology and clinical laboratory testing, to support patient care in hospitals, clinics, and other healthcare settings. Robert Baker, Operations Manager of the molecular hematology lab at HSL, provides comprehensive molecular testing services, including the use of NGS for a broad range of hematologic malignancies. The lab processes hundreds of hematological samples annually. Previously, the lab relied on a combination of PCR assays and NGS for molecular analysis. However, this workflow, while effective, had limitations in terms of labor intensity and throughput.
Prior to adopting Rapid NGS testing, HSL relied on multiple single-analyte tests along with a traditional NGS assay. Comprehensive results for all biomarkers generally takes about two weeks.
While this method allowed for rapid initial detection, it was labor-intensive, had low throughput, and often required subsequent NGS to uncover additional significant variants.
In response to the need for a more efficient approach, Baker’s team conducted a feasibility study using the Ion Torrent Genexus System with the Oncomine Myeloid Assay GX v2. This system enables rapid turnaround times and comprehensive genomic profiling. By pooling samples and optimizing the workflow, the lab could process up to 2,500 samples annually on a single sequencer, significantly enhancing their capacity.
Key features of the new workflow
The new workflow demonstrated several advantages:
Building on the success with myeloid malignancies, the laboratory also utilized the Ion AmpliSeq Liverpool Lymphoid Network Panel for rapid genomic profiling of lymphoid malignancies. This panel enabled the laboratory to quickly obtain genomic data within approximately 27 hours, effectively supporting the analysis of low-grade T- and B-cell lymphoma samples. Additionally, the laboratory leveraged a network of laboratories for ongoing analytical validation and updates, enabling the reliability and accuracy of the profiling results.
HSL has also started to explore new NGS applications for B-cell lymphomas. They are evaluating the use of gene expression signatures as a rapid method for detecting single/double/triple hit high-grade B-cell lymphomas.
Mr. Baker presented preliminary data from an AML sample profiled using the Oncomine Myeloid MRD (RUO) Assay and Ion GeneStudio S5 system. This assay uses AmpliSeq™HD technology to enable a low Limit of Detection (LoD) down to 0.05% variant allele frequency (VAF) for studying measurable residual disease (MRD). In their initial run, the lab was able to detect a low-frequency DNMT3A variant at 0.14% VAF and an IDH2 variant at 0.07%VAF.
Mr. Baker presented data suggesting that overexpression in MYC, BCL2, BCL6 may be a reliable indicator of fusion involving the overexpressed gene(s). In their workflow, a gene expression profile can be rapidly measured using the Oncomine Childhood Cancer Research Assay (OCCRA). Any samples with an overexpression of MYC, BCL2, or BCL6 are then tested using an orthogonal NGS fusion assay to confirm the presence of a translocation.This approach may result in a fast and efficient way to perform a primary analysis without the need to run a separate NGS fusion assay, though Mr. Baker stressed that more research must be done in this area.
Some potential future strategies for lymphoma genomic profiling include subjecting all high-grade lymphomas to both the OCCRA and Liverpool Lymphoid Network Panels, while low-grade lymphomas would only be analyzed using the Liverpool Lymphoid Network Panel.
The advancements presented by Robert Baker highlight the transformative impact of rapid NGS technologies in the field of hematological malignancies. By significantly reducing turnaround times and improving genomic profiling accuracy, these innovations may help enhance future clinical decision-making and streamline laboratory workflows.
We will be happy to answer your questions and provide a demo of our NGS solutions.
For Research Use Only. Not for use in diagnostic procedures.
CN: 61355