Thermo Fisher Scientific is an industry-leading, trusted partner for pathogen detection research solutions that are critical to help combat the current SARS-CoV-2 crisis. We have expanded our current portfolio of SARS-CoV-2 research solutions to include new sensitive, multiplex, real-time PCR kits specifically designed for pathogen detection research during the overlap of the SARS-CoV-2 crisis and respiratory virus season.
For Research Use Only. Not for use in diagnostic procedures.
For the most up-to-date information on the current SARS-CoV-2 outbreak, visit the WHO or CDC websites.
Multiplex kit for SARS-CoV-2, influenza A, and influenza B (For Research Use Only)
- Multiplex detection of RNA from SARS-CoV-2 (S & N genes), influenza A, and influenza B
- Designed using two SARS-CoV-2 targets to compensate for emerging variants and mutations, providing confidence in results
- Kit delivers 1,000 reactions
- Recommended instruments: 7500 Fast Dx and QuantStudio 5 (96-well, 0.2 mL) real-time PCR systems
Multiplex kit for SARS-CoV-2, influenza A/B, and respiratory syncytial virus (RSV) (For Research Use Only)
- Multiplex detection of RNA from SARS-CoV-2 (S & N genes), influenza A/B, and RSV A/B
- Designed using two SARS-CoV-2 targets to compensate for emerging variants and mutations, providing confidence in results
- Kit delivers 1,000 reactions
- Recommended instrument: 7500 Fast Real-Time PCR System
Product | Description | Quantity | Cat. no. |
---|---|---|---|
Multiplex solution for SARS-CoV-2, influenza A, and influenza B | |||
TaqMan SARS-CoV-2, Flu A, Flu B RT-PCR Assay Kit | SARS-CoV-2 (spike (S) and nucleocapsid (N) gene), Flu A, and Flu B multiplex; and MS2 phage control | 1,000 reactions | A47701 |
TaqMan SARS-CoV-2, Flu A, Flu B RNA Control | RNA control: SARS-CoV-2 (S and N gene), Flu A, Flu B | 10 x 10 µL | 956125 |
TaqMan SARS-CoV-2 Control Dilution Buffer | Dilution buffer for all RNA controls | 10 x 250 µL | A49889 |
TaqPath 1-Step RT-qPCR Master Mix | 4X RT-qPCR master mix for up to four different RNA/DNA targets in a single multiplex reaction | 1 x 10 mL | A28523 |
Multiplex solution for SARS-CoV-2, influenza A/B, RSV A/B | |||
TaqMan SARS-CoV-2, Flu A/B, RSV RT-PCR Assay Kit | SARS-CoV-2 (S and N gene), Flu A/B, and RSV A/B multiplex; and MS2 phage control | 1,000 reactions | A47702 |
TaqMan SARS-CoV-2, Flu A/B, RSV RNA Control | RNA control: SARS-CoV-2 (S and N gene), Flu A/B, RSV | 10 x 10 µL | 956126 |
TaqMan SARS-CoV-2 Control Dilution Buffer | Dilution buffer for all RNA controls | 10 x 250 µL | A49889 |
TaqPath 1-Step RT-qPCR Master Mix | 4X RT-qPCR master mix for up to four different RNA/DNA targets in a single multiplex reaction | 1 x 10 mL | A28523 |
Recommended analysis software
Design and Analysis Software Version 2.5.0
Recommended qPCR systems for use with the above multiplex kits
7500 Fast and 7500 Fast Dx Real-Time PCR systems
Applied Biosystems 7500 Fast and 7500 Fast Dx Real-Time PCR systems offer high performance, multicolor real-time PCR wherever your research takes you. Our 7500 Fast (0.1 mL) system is designed to deliver the reliable performance and dye flexibility you need to generate data that you can trust.
QuantStudio 5 Real-Time PCR System (0.2-mL 96-well block)
The Applied Biosystems QuantStudio 5 Real-Time PCR System is ideal for all levels of users who need a high-performance instrument with features for maximum control of their experiments. Includes built-in security features, locked workflows, user account management, and options for web browser–based or desktop analysis.
Viral nucleic acid isolation products
MagMAX Viral/Pathogen II Nucleic Acid Isolation Kit
Isolate high-quality DNA and RNA quickly and efficiently using the Applied Biosystems MagMAX Viral/Pathogen II Nucleic Acid Isolation Kit.
- Automation-ready protocols designed for the KingFisher Flex system
- Fast procedure allows for 96 samples to be processed in <30 minutes
- Flexible protocol accommodates research sample volume inputs from 200 to 400 µL
- Elution volumes ranging from 50 to 100 µL
KingFisher Flex system
Automate your DNA and RNA isolation with the KingFisher Flex system, a versatile benchtop instrument that processes 24 or 96 samples per run.
- High-throughput purification of 24 or 96 samples per run
- Easy to install and run (ready to run in 10 minutes or less)
- Choose from two plate formats for wide volume range (20–5,000 µL)
- Download existing protocols from library, or easily customize protocols using Thermo Scientific BindIt Software
For Research Use Only. Not for use in diagnostic procedures.