Post-Transplant Monitoring
Precision-driven post-transplant diagnostic tests
As the trusted partner of HLA labs around the world, the Transplant Diagnostics business of Thermo Fisher Scientific provides essential assays and services that deliver reliable and actionable data.
Expand your lab's capabilities with our post-transplant monitoring assays and services
Antibody monitoring assays
Molecular monitoring assays
Gene expression profiling services
Explore our post-transplant monitoring solutions
Antibody monitoring assays
Precise HLA antibody detection, key to transplant success. Detect HLA antibodies, especially donor-specific ones (DSAs), linked to transplant rejection. Pregnancy, transfusions, or mismatched grafts can trigger antibody formation. Early DSA detection and intervention improve outcomes. Our reliable, high-sensitivity single-bead assays offer optimal antibody monitoring for better transplant care.
Cell-free DNA monitoring assay
Accept cfDNA is an NGS assay designed to detect and analyze circulating dd-cfDNA in post-transplant human plasma samples. Its unique indel design allows for accurate results, reduces amplification and sequencing errors. Combined with an efficient workflow, Accept cfDNA is the ideal tool for post-transplant monitoring and provides early time to result when run in your own in house HLA lab.
Chimerism monitoring assay
Designed to provide accurate and sensitive detection of microchimerism levels in stem cell transplant samples, the One Lambda Devyser Chimerism NGS assay is a post-transplant monitoring solution that is perfect for HLA and molecular core labs associated with HSCT Programs.
Gene expression profiling services
MMDx Heart and MMDx Kidney complement conventional biopsy processing to improve the assessment of rejection and injury in transplanted organs. These Laboratory Developed Tests use a GeneChip™ Custom Microarray to measure mRNA transcript levels in biopsies along with extensive big data derived from individuals and populations.
Not all products are CE marked or have 510(k) clearance for sale in the U.S. Availability of products in each country depends on local regulatory marketing authorization status.