Search Thermo Fisher Scientific
The TypeStream Visual NGS Analysis Software (TSV) is a stand-alone NGS software solution that supports analysis of both single-read and paired-end sequencing data produced using the One Lambda AllType NGS Assays.
Upon completion of sequencing, the TypeStream Visual software automatically imports and aligns unmapped barcode sequences to the IMGT™ reference library to generate allele assignments. In addition to genotyping, the TypeStream Visual interface provides a wide range of analytical tools, run statistics, and quality metrics.
Data for One Lambda HybriType assay is analyzed using our NGS software – TypeStream Visual providing automated and streamlined analysis with a wide range of analytical tools, run statistics and quality metrics.
Copy Number Variation (CNV) tool provides information on the number of copies of each allele per locus. TypeStream Visual analyzes read counts by comparing within and across samples:
A complete list of features and version enhancements can be found in the respective Release Notes located on the TSV product support site.
TypeStream Visual software includes bi-directional communication with HistoTrac, enabling the seamless transfer of results between both systems for better data management, reduced transcription errors, and optimized reporting. Results may be combined in our HLA Fusion Software for advanced analytics.
Ensure you are upgraded to the latest version of TSV to benefit from new functionality and enhancements, ensuring the most favorable experience in analysis performance.
Not all products are CE marked or have 510(k) clearance for sale in the US. Availability of product in each country depends on local regulatory marketing authorization status. Please consult your local sales representative for details.