500
Analyze 500 TaqMan® Array Cards
10 x
Up to 10x
Faster Analysis
100
Analyze 100 OpenArray® Plates
Applied Biosystems™ qPCR analysis modules/apps allow multi-plate analysis of real-time PCR data with unprecedented ease of use and improved capacity and performance. Analysis of hundreds of 384-well plates can be achieved in just minutes. Applied Biosystems qPCR software is compatible with experiment files from the Applied Biosystems QuantStudio™ 12K Flex, QuantStudio™ 7 Flex, QuantStudio™ 6 Flex, QuantStudio™ 3, QuantStudio™ 5, 7900HT Fast, 7500/7500 Fast, and StepOne™/StepOnePlus™ Real-Time PCR Systems.
Fast and powerful data analysis for your genomics platforms
Applied Biosystems analysis modules are innovative cloud-based secondary data analysis solutions that bring together multiple data sets in one convenient place. This online solution makes it easier to view, store, and analyze qPCR and Sanger sequencing data. Applied Biosystems analysis modules take advantage of cloud computing to provide highly versatile analysis tools that are flexible, fast, and easy to use.
Applied Biosystems qPCR Analysis Modules are compatible with experiment files from any Applied Biosystems instrument including the most recent additions to the QuantStudio family such as the QuantStudio 3 and QuantStudio 5 Real-Time PCR Systems, as well as the QuantStudio 6 Flex System, QuantStudio 7 Flex System and QuantStudio 12k Flex System.
Find out more about the QuantStudio family of qPCR instruments
Applications include:
  • Absolute quantification
  • Gene expression
  • Genotyping
  • High Resolution Melt (HRM)
  • Presence/Absence
Get started using these modules/apps by first accessing Thermo Fisher Cloud.
Don't have an account? Start here ›
Introducing the Applied Biosystems analysis modules for qPCR and Sanger sequencing
UX DATA-TRKR : Contact Global UX for removal