47b9 - Apps/Techniques/Products/Media Gallery/Contact Us
Electron Microscopy
Bio-pharmaceutical Research with Electron Microscopy
Bio-pharma drug discovery through high resolution cryo-electron microscopy.
Rather than starting with a disease and searching for its origin, protein-based biopharmaceuticals (biologics) begins with the detection of a genetic variation and then relies on therapies that manipulate it. Cryo-EM helps researchers examine a broad range of biological structures and provides the ability to visualize otherwise difficult or complicated biologics.
For example, protein samples, which are frozen in vitreous (non-crystalline) ice, can be examined in a near-native, fully hydrated state, thereby enabling accelerated structural drug discovery. Near-atomic-resolution cryo-electron microscopy can be employed throughout the drug discovery process, from target identification, through lead characterization, to epitope mapping.
Media Gallery
Applications
Life_Sciences_Application_274x180_144DPI
Structural Biology Research
Cryo-electron microscopy enables the structural analysis of challenging biological targets such as large complexes, flexible species and membrane protein.
Infectious_Disease_Research_App_w274x180
Infectious Disease Research
Cryo-EM techniques enable multiscale observations of 3D biological structures in their near-native states, informing faster, more efficient development of therapeutics.
DrugDiscovery_Appl_274x180
Drug Discovery
Learn how to take advantage of rational drug design for many more major drug target classes, leading to best-in-class drugs.
Techniques
Single Particle Analysis
Single particle analysis (SPA) is a cryo-electron microscopy technique that enables structural characterization at near-atomic resolutions, unraveling dynamic biological processes and the structure of biomolecular complexes/assemblies.
Learn more ›
Cryo-Tomography
Cryo-electron tomography (cryo-ET) delivers both structural information about individual proteins as well as their spatial arrangements within the cell. This makes it a truly unique technique and also explains why the method has such an enormous potential for cell biology. Cryo-ET can bridge the gap between light microscopy and near-atomic-resolution techniques like single-particle analysis.
Learn more ›
MicroED
MicroED is an exciting new technique with applications in the structural determination of small molecules and protein. With this method, atomic details can be extracted from individual nanocrystals (<200 nm in size), even in a heterogeneous mixture.
Learn more ›
Single Particle Analysis
Single particle analysis (SPA) is a cryo-electron microscopy technique that enables structural characterization at near-atomic resolutions, unraveling dynamic biological processes and the structure of biomolecular complexes/assemblies.
Learn more ›
Cryo-Tomography
Cryo-electron tomography (cryo-ET) delivers both structural information about individual proteins as well as their spatial arrangements within the cell. This makes it a truly unique technique and also explains why the method has such an enormous potential for cell biology. Cryo-ET can bridge the gap between light microscopy and near-atomic-resolution techniques like single-particle analysis.
Learn more ›
MicroED
MicroED is an exciting new technique with applications in the structural determination of small molecules and protein. With this method, atomic details can be extracted from individual nanocrystals (<200 nm in size), even in a heterogeneous mixture.
Learn more ›
Products
Style Sheet for Instrument Cards Original
Krios Rx Cryo-TEM
  • Industry leading productivity and ease of use
  • Fixed 300 kV voltage for SPA only
  • Pharma-dedicated service package for guaranteed reliability
Krios G4 Cryo-TEM
  • Improved ergonomics
  • Fits more easily into new and existing labs
  • Maximized productivity and automation
  • Best image quality for high-resolution 3D reconstruction
Glacios Cryo-TEM
  • Flexible Accelerating Voltage 80-200 kV
  • Industry-leading Autoloader for cryogenic sample manipulation
  • Small footprint
  • Enhanced ease-of-use
Tundra Cryo-TEM
  • Structural information at biologically relevant resolution
  • Space efficient and cost effective
  • Easy, iterative sample optimization
  • Unique AI algorithms for streamlined data collection
Talos Arctica TEM
  • Increased data acquisition speed
  • High data with robotic sample handling & automated loading
  • Unattended platform operation and automated data acquisition
  • Low cost of ownership with remote diagnostics and preventive service
Talos F200C TEM
  • Flexible EDS analysis reveals chemical information
  • High-contrast, high-quality TEM and STEM imaging
  • Ceta 16 Mpixel CMOS camera provides large field of view and high read-out speed
Talos L120C TEM
  • Increased stability
  • 4k × 4K Ceta CMOS camera
  • TEM magnification range from 25–650 kX
  • Flexible EDS analysis reveals chemical information
Prisma E SEM
  • Entry-level SEM with excellent image quality
  • Easy and quick sample loading and navigation for multiple samples
  • Compatible with a wide range of materials thanks to dedicated vacuum modes
Quattro ESEM
  • Ultra-versatile high-resolution FEG SEM with unique environmental capability (ESEM)
  • Observe all information from all samples with simultaneous SE and BSE imaging in every mode of operation
Apreo 2 SEM
  • High-performance SEM for all-round nanometer or sub-nanometer resolution
  • In-column T1 backscatter detector for sensitive, TV-rate materials contrast
  • Excellent performance at long working distance (10 mm)
Verios 5 XHR SEM
  • Monochromated SEM for sub-nanometer resolution over the full 1 keV to 30 keV energy range
  • Easy access to beam landing energies as low as 20 eV
  • Excellent stability with piezo stage as standard
VolumeScope 2 SEM
  • Isotropic 3D data from large volumes
  • High contrast and resolution in high and low vacuum modes
  • Simple switch between normal SEM use and serial block-face imaging
Helios Hydra DualBeam
  • 4 fast switchable ion species (Xe, Ar, O, N) for optimized PFIB processing of a widest range of materials
  • Ga-free TEM sample preparation
  • Extreme high resolution SEM imaging
Scios 2 DualBeam
  • Full support of magnetic and non-conductive samples
  • High throughput subsurface and 3D characterization
  • Advanced ease of use and automation capabilities
Aquilos 2 Cryo-FIB
  • Automation enables production of multiple lamellas
  • Target and extract your structure of interest with lift-out nano-manipulator
  • 3D visualization for high-resolution tomography
Vitrobot System
  • Fully Automated Sample vitrification
  • Blotting Device
  • Semi-Automated Grid Transfer
  • High Sample Throughput
Selectris and Selectris X Imaging Filters
  • Designed for high stability and atomic resolution imaging
  • Straightforward operation
  • Paired with the latest generation Thermo Scientific Falcon 4 Direct Electron Detector
Ceta-D Camera
  • Optimum performance at any high tension (20–300 kV)
  • Compatible with post-column filters and spectrometers
  • Movie acquisition for dynamic studies
Falcon 4 Detector
  • Leading detective quantum efficiency
  • 10x shorter exposure time than its predecessor
  • Fully embedded in Thermo Scientific software
  • Built in data management
Tomography 5
Software
  • Automated multi-site batch tomography
  • Automatic cassette mapping
  • Dose symmetric tilt scheme for optimal electron dose
  • Fully integrated with Selectris Imaging Filters
Maps Software
  • Acquire high resolution images over large areas
  • Easily find regions of interest
  • Automate image acquisition process
  • Correlate data from different sources
Auto Slice and View 4.0 Software
  • Automated serial sectioning for DualBeam
  • Multi-modal data acquisition (SEM, EDS, EBSD)
  • On-the-fly editing capabilities
  • Edge based cut placement
Inspect 3D Software
  • Image processing tools and filters for cross-correlation
  • Feature tracking for image alignment
  • Algebraic reconstruction technique for iterative projection comparison
Amira Software
Life Sciences
  • Import and process image data
  • Auto-segmentation, object separation, labeling
  • Analyze and quantify with automated statistics
  • Create images, animations and videos
EPU 2 Software
  • Microscope-embedded solution for single particle acquisition
  • Optimized for high-throughput particle collection
  • Compatible with film, CCD cameras, and direct electron detectors
Contact us
Style Sheet to change H2 style to p with em-h2-header class
Style Sheet to change H3 to p with em-h3-header class
Style Sheet to change Applications H3 to p with em-h3-header class
Style Sheet for Komodo Tabs
Style Sheet for Support and Service footer
Style Sheet for Fonts
Style Sheet for Cards