Faster forensic gunshot residue analysis that you can truly trust
Above all, gunshot residue (GSR) analysis needs to be utterly reliable. But it also needs to be easy and deliver lightning-fast turnaround.
While scanning electron microscope (SEM) technology is well established in GSR analysis, the Thermo Scientific Phenom ParticleX GSR Desktop SEM was custom designed for the task.
It delivers compositional analysis at the micro level that helps you find every particle. And it’s fast, making it possible to analyze a sample that might take conventional SEMs four hours in as little as 15 minutes.
Featuring integrated automation with Thermo Scientific Perception Software, the Phenom ParticleX GSR Desktop SEM can also help you analyze a greater quantity of samples with higher quality, delivering concise industrial reports that turn raw data into clear and unbiased answers for you and your team.
This system’s integrated energy-dispersive X-ray spectroscopy (EDS) accurately analyzes the compositional elements of the particles on a GSR sample to identify whether gunshot residue is present via one simple process. 
Phenom ParticleX GSR Desktop SEM overview
Utterly reliable results
Every particle counts in GSR analysis. The Phenom ParticleX GSR Desktop SEM gives you round-the-clock, in-depth analysis that you can truly trust to identify the smallest GSR particles.  
A super-speedy SEM
The Phenom ParticleX GSR Desktop SEM maintains high quality while drastically accelerating your throughput—and your return on investment.
Automation makes things easy
Perception Software can help you work smarter and faster—with minimum intervention. For example, internal scan control enables more accurate beam positioning, which is key when revisiting particles in the GSR verification phase. 
Extensive elemental analysis
Integrated EDS helps you analyze the elemental composition of GSR powder, which is essential for forensic lab teams.
Phenom ParticleX GSR Desktop SEM resources
Would you like to know more about this system? Dive into a wealth of resources, from eBooks to datasheets. 
Particle analysis learning center
Learn more about the role of electron microscopy in particle analysis with our collection of articles, eBooks, and webinars.
For Research Use Only. Not for use in diagnostic procedures.