High-performance imaging with integrated chemical analysis and structural characterization
The new Thermo Scientific Apreo ChemiSEM System supports your materials science research by simplifying the imaging process, making it accessible to both expert users and newcomers alike. With innovative features like Smart Frame Integration (SFI) and newly developed autofocus and autostigmation functions, it can help you easily obtain clear, high-resolution images as well as results for elemental analysis (EDS) and diffraction (EBSD).
Thermo Scientific ChemiSEM Technology integrates energy dispersive spectroscopy (EDS) capabilities directly into the microscope's user interface, which simplifies analysis and helps deliver reliable results with minimal user bias. With live quantification algorithms and automated data processing, ChemiSEM Technology delivers speed and accuracy to help you easily uncover hidden features and gain a more comprehensive understanding of your materials.
Apreo ChemiSEM System overview
SEM imaging for every user
The new Smart Frame Integration (SFI) helps you easily improve your imaging results and the newly developed autofocus and autostigmation functions help beginners enhance imaging performance.
Complete material analysis
ChemiSEM Technology and dedicated EBSD Software make advanced analyses more accessible. The new enhanced TruePix Electron Backscatter Diffraction (EBSD) Detector and EBSD Software guides you through every step of the workflow, from defining settings to collecting and processing results.
No need for additional electron detectors
The Apreo ChemiSEM System's Trinity Detection System can help you gather accurate results without compromise and delivers exceptional resolution and signal filtering options. It can also be paired with Everhart-Thornley, retractable backscattered, cathodoluminescence, and STEM detectors.
Unlock the full potential of your materials analysis research
Whether you're a seasoned researcher or new to electron microscopy, the Apreo ChemiSEM System is your gateway to a world of discovery. Its customizable interface, advanced features, and performance ensure that high-quality imaging and analyses are within reach for every user, in every environment.
Apreo ChemiSEM System videos
See the Apreo ChemiSEM System in action and see what it has to offer. 
Apreo ChemiSEM System resources
Find more information about the Apreo ChemiSEM System in our eBooks, datasheets, and other resources.
For Research Use Only. Not for use in diagnostic procedures.