Multiple-mode, high-resolution environmental SEM
With its range of vacuum levels, the Thermo Scientific Prisma E SEM helps you analyze a huge range of materials in high resolution, in their natural state and with complete confidence. This includes wet, hot, dirty, and outgassing samples. Also featured is through-the-lens differential pumping and simultaneous secondary electron and backscattered electron imaging in all modes.
The Prisma E SEM allows you to easily set up the acquisition for a wide range of materials such as cement, solar cells, or geological materials that are commonly known to be outgassing and beam-sensitive.
Faster, smarter materials analysis with the Prisma E SEM
In addition to expanding the scope of your experiments, the Prisma E SEM enables you to achieve this  quickly and efficiently. A simple interface (with integrated user guidance), coupled with built-in automation, make this system simple to learn. And if mistakes are made, just use its undo function.
Integrated EDS analysis for elevated elemental characterization
The Prisma E SEM makes elemental analysis  intuitive and efficient. The integrated EDS capability provides always-on analytical capability, with no time-consuming switching between systems.  The Prisma E SEM offers an easy, user-friendly way to obtain live composition-based image coloring.
Integrated EDS analysis with ESEM on the Prisma E SEM for live composition-based image coloring.
Built for your biggest analytical needs
For scientists looking to step up the breadth and depth of their analytical research, perhaps through the ability to analyze beam-sensitive samples, it can be challenging to find an economical solution that ticks all the boxes. The Prisma E SEM was created to do just that. It gives you remarkable freedom and flexibility—all with fewer mouse clicks.
Prisma E SEM technical specifications
Sample size
  • Up to 122 mm diameter
  • Up to 50 mm height
Source type Tungsten (W)
Max. resolution  3 nm
Detectors and signals
  • Everhart-Thornley (ETD) SE detector (standard)
  • Low-vacuum SE detector (LVD) (standard)
  • Gaseous SED (GSED) (standard)
  • Retractable backscattered electron detector (optional)
  • Energy-dispersive X-ray (EDS) detector (optional)
  • Low-vacuum gaseous analytical backscattered electron detector (optional)
  • Cathodoluminescence (CL) detector (optional)
  • Retractable STEM detector (optional)
  • WetSTEM detector (optional)
Software options and accessories
  • ColorSEM Technology
  • Maps 3 Software
  • AutoScript 4 Software
  • Avizo Software
  • Avizo Trueput Software
Footprint 890(w) x 1268 (d) x 1640 (h) mm, 550 kg
Acceleration voltage range  200 V–30 kV
Low vacuum mode (stating pressure ranges) Up to 130 Pa
For Research Use Only. Not for use in diagnostic procedures.