Search Thermo Fisher Scientific
Thermo Scientific Spectra Transmission Electron Microscopes are compatible with a wide range of software to help you streamline your workflows, more accurately interpret your data, and shorten your learning curve.
Unlock the full potential of your transmission electron microscopy experiments with Thermo Scientific Velox Software. This advanced program delivers exceptional control, reproducibility, and support for quantitative (S)TEM and TEM material analysis. With its advanced EDS packages and integration with Super-X and Dual-X Detector Systems, Velox Software helps to ensure high-yield acquisition of outstanding EDS spectrum images. Featuring a user-friendly interface, drift compensation methods, and time resolved XEDS capabilities, Velox Software enables precise compositional mapping and efficient results.
Thermo Scientific AutoScript Software allows you to create automated data analysis, customizable workflows, and seamless integration with your microscope, helping you achieve faster, more accurate results.
Thermo Scientific Maps Software is an imaging and correlative workflow software suite designed for SEM, FIB-SEM, and TEM platforms. It provides a powerful and easy-to-use platform for automating imaging workflows and preserving the context of observations. With Maps Software, you can perform multi-scale imaging automation, correlative microscopy, integrated analytics, intuitive visualization, and offline collaboration. The software also integrates with Thermo Scientific Avizo Software for image processing and statistical data generation.
Thermo Scientific Tomography 5 Software brings you the latest advances in EM tomography, allowing you to acquire valuable 3D data, even on your most challenging samples. Its new user-friendly interface enables both experienced and new users to jumpstart their tomography data acquisition.
For Research Use Only. Not for use in diagnostic procedures.