c7c8 - Key/Specs/Resources/Appl/Tech/Doc/Contact
The Thermo Scientific Talos Arctica Cryo-TEM is a powerful, stable, and versatile 200kV FEG scanning and transmission electron microscope (S/TEM) built for delivering high-resolution 3D characterization of biological samples and biomaterials in cell biology, structural biology, and nanotechnology research. The Talos Arctica S/TEM enables scientists to quickly obtain better insight and understanding of macromolecular structures, cellular components, cells, and tissues in three dimensions.
Transmission Electron Microscopes
Talos Arctica Cryo-TEM for Life Sciences
3D cryo TEM analysis tool for biological samples and biomaterials.
Key Features
Fast 200 kV FEG S/TEM for sample analysis in multiple dimensions
The Talos Arctica Cryo-TEM is built to deliver rapid access to 2D and 3D data so that you can concentrate on discovery. The system's constant-power, C-Twin lens has outstanding optical performance, ensuring an optimal balance of contrast and resolution. Additionally, the ultra-stable platform includes a piezo stage with stable electronics and a rugged system enclosure for maximum thermal and mechanical stability.
Increased data acquisition speed
Acquisition speeds have been improved up to 3 times for tomography and up to 1.5 times for single particle analysis (with Thermo Scientific EPU Software) compared to side entry loading systems.
High data throughput
Robotic sample handling and automated loading for up to 12 samples provides up to 40% enhancement in data throughput.
Unattended operation
Auto-filling of liquid nitrogen and powerful applications software allows for continuous unattended platform operation and automated data acquisition.
Low cost of ownership
New design allows for remote diagnostics and preventive service, lowering ownership cost and maximizing systems uptime.
Specifications
Style Sheet for Products Table Specifications
Source
  • High-brightness field emission gun (X-FEG)
  • Accelerating voltage
  • 80–200 kV
  • Cryo-Autoloader
  • Automated and contamination-free loading of cassettes, containing up to 12 Autogrids
  • Software
  • EPU Software for SPA screening and data acquisition
  • Temperature management software – Includes liquid nitrogen autofill and cool down scheduling
  • Low-dose software suite minimizes electron dose during cryo-TEM operation
  • Lenses
  • Automatic condenser, objective and SA apertures
  • Symmetric constant power objective lens (minimize image aberrations and lens hysteresis during mode switching)
  • Stages
  • Computerized 4-axes specimen stage with ±70° alpha tilt
  • Cryo-stage with single axis holder
  • Imaging
  • Rotation-free imaging upon magnification changes
  • Pole piece gap
  • 11 mm
  • Structural components
  • Two 24” monitors to be placed within 10 m of the column
  • Cameras
  • Digital FluCam – All manual and automatic alignments can be executed with the search and view camera
  • Ceta 16M Camera
  • Installation requirement highlights
  • Environmental temperature: 18°C – 23°C
  • Size: 5765 x 3895 x 3030
  • Detectors (optional)
  • Falcon 3EC Direct Electron Detector
  • HAADF STEM detector
  • On-axis BF/DF detectors
  • Software (optional)
  • STEM and TEM tomography software
  • Service package
  • Accelerate integrated service and application support packages
  • Other options
  • Volta phase plate 
  • compatible with Gatan post column imaging filter
  • Operating system
  • Windows 10
  • Resources
    Applications
    Infectious disease research
    Infectious Disease Research
    Cryo-EM techniques enable multiscale observations of 3D biological structures in their near-native states, informing faster, more efficient development of therapeutics.
    Drug discovery
    Drug Discovery
    Learn how to take advantage of rational drug design for many major drug target classes, leading to best-in-class drugs.
    Structural biology research
    Structural Biology Research
    Cryo-electron microscopy enables the structural analysis of challenging biological targets such as large complexes, flexible species and membrane protein.
    Techniques
    Single Particle Analysis
    Single particle analysis (SPA) is a cryo-electron microscopy technique that enables structural characterization at near-atomic resolutions, unraveling dynamic biological processes and the structure of biomolecular complexes/assemblies.
    Learn more ›
    Cryo-Tomography
    Cryo-electron tomography (cryo-ET) delivers both structural information about individual proteins as well as their spatial arrangements within the cell. This makes it a truly unique technique and also explains why the method has such an enormous potential for cell biology. Cryo-ET can bridge the gap between light microscopy and near-atomic-resolution techniques like single-particle analysis.
    Learn more ›
    MicroED
    MicroED is an exciting new technique with applications in the structural determination of small molecules and protein. With this method, atomic details can be extracted from individual nanocrystals (<200 nm in size), even in a heterogeneous mixture.
    Learn more ›
    Automated Particle Workflow
    The Automated NanoParticle Workflow (APW) is a transmission electron microscope workflow for nanoparticle analysis, offering large area, high resolution imaging and data acquisition at the nanoscale, with on-the-fly processing.
    Learn more ›
    Single Particle Analysis
    Single particle analysis (SPA) is a cryo-electron microscopy technique that enables structural characterization at near-atomic resolutions, unraveling dynamic biological processes and the structure of biomolecular complexes/assemblies.
    Learn more ›
    Cryo-Tomography
    Cryo-electron tomography (cryo-ET) delivers both structural information about individual proteins as well as their spatial arrangements within the cell. This makes it a truly unique technique and also explains why the method has such an enormous potential for cell biology. Cryo-ET can bridge the gap between light microscopy and near-atomic-resolution techniques like single-particle analysis.
    Learn more ›
    MicroED
    MicroED is an exciting new technique with applications in the structural determination of small molecules and protein. With this method, atomic details can be extracted from individual nanocrystals (<200 nm in size), even in a heterogeneous mixture.
    Learn more ›
    Automated Particle Workflow
    The Automated NanoParticle Workflow (APW) is a transmission electron microscope workflow for nanoparticle analysis, offering large area, high resolution imaging and data acquisition at the nanoscale, with on-the-fly processing.
    Learn more ›
    Contact us
    Style Sheet to change Applications H3 to p with em-h3-header class
    Style Sheet to change H3 to p with em-h3-header class
    Style Sheet to change H2 style to p with em-h2-header class
    Style Sheet for Komodo Tabs
    Electron microscopy services for
    the life sciences
    To ensure optimal system performance, we provide you access to a world-class network of field service experts, technical support, and certified spare parts.
    Style Sheet for Support and Service footer
    Style Sheet for Fonts
    Style Sheet for Cards