usda-showcase-230x195-r3
Help improve testing efficiency and reliability with our USDA-licensed diagnostic solutions
The USDA Animal and Plant Health Inspection Service (APHIS) regulates veterinary biologics (vaccines, bacterins, antisera, diagnostic kits, and other products of biological origin) to help ensure that the veterinary biologics available for the diagnosis, prevention, and treatment of animal diseases are safe and effective. The APHIS Center for Veterinary Biologics (CVB) carries out this work, focusing on enforcement of the Virus Serum Toxin Act.
Thermo Fisher Scientific animal health products have been at the forefront of recent advances in production animal diagnostics, and are some of the leading modern diagnostic tools to veterinary laboratories around the world.
In keeping with our commitment to deliver the highest-quality next-generation molecular tools, we provide several farm animal diagnostic solutions that have been validated through the stringent USDA licensure process. This includes innovative USDA-licensed detection kits that offer rapid, easy-to-use PCR-based testing solutions that are designed to detect bovine viral diarrhea virus (BVDV), swine influenza virus (SIV), and avian influenza virus (AIV) with exceptional specificity, sensitivity, and reproducibility.
Our vision is to provide comprehensive solutions to help diagnose farm animal health issues more quickly and accurately than ever before. To learn more about our USDA-licensed diagnostic solutions for such diseases as tritrichomonas foetus, bovine viral diarrhea virus and swine influenza virus please download our product flyer.
  View product flyer ›
Demystifying USDA licensure
Thermo Fisher Scientific hosted an event attended by state veterinarians that included a presentation explaining the USDA licensure process, as well as the production and performance criteria that veterinary diagnostic test kits must satisfy to earn a USDA license. See below to view the presentation video.
Mary Anne Williams
Senior Manager, Regulatory Affairs
Thermo Fisher Scientific
Watch it now ›
Free webinar
Register for “Demystifying USDA licensure and its importance to diagnostics testing”. Please click the "Register" button below to attend this free ondemand webinar and learn about the process to obtain a USDA license, the importance of using USDA-licensed kits, and the benefits to laboratories, veterinarians, and health officials.
Register for webinar
VetMAX-Gold USDA-licensed solutions
Applied Biosystems™ VetMAX™-Gold Trich Detection Kit—The only USDA-licensed, real-time PCR test for the rapid and accurate detection of Tritrichomonas foetus (T. foetus) in bulls.
  • Sensitive, qualitative real-time PCR assay yielded up to 100% sensitivity in field trials
  • Detection kit can be used to test pools of up to 5 samples with 96.16% accuracy
  • Helps improve reproducibility across laboratories and across states
  • Simplifies lab workflow and is easily automated for high-throughput processing
  • USDA oversees the quality control process and the release of every batch
Find out more ›    Download product flyer ›
Applied Biosystems™ VetMAX™-Gold BVDV PI Detection Kit—USDA-licensed, real-time PCR test for the detection of bovine viral diarrhea virus (BVDV) persistent infection (PI) in cattle.
  • Approved for use with single samples or pooled samples of up to 24 animals
  • Proven repeatability (CV <3.3%) and reproducibility (CV <2.4%) in field studies
  • Rapid and cost-effective method for detection of BVDV, with results in ~1.5 hr
  • Workflow minimizes sample handling and helps reduce sample cross-contamination
  • USDA oversees the quality control process and the release of every batch
Find out more ›    Download product flyer ›
Applied Biosystems™ VetMAX™-Gold MAP Detection Kit—A USDA-licensed real-time PCR test for the detection of Mycobacterium avium subsp. paratuberculosis (MAP) in beef and dairy cattle.
  • Validated for use with single cattle feces samples or pooled samples of up to 5 animals
  • Demonstrated high sensitivity (96.2%) and specificity (96.4–100%) in USDA validation study
  • Proven repeatability (100%) and reproducibility (100%) in USDA validation study
  • Simple workflow tailored for MAP detection that is easily automated for high-throughput processing
  • Includes Applied Biosystems™ VetMAX™ Xeno™ Internal Positive Control to help prevent false signals from nonspecific targets
  • Routine monitoring of our manufacturing site for compliance with production and quality system guidelines
Find out more ›           Download product flyer ›
Applied Biosystems™ VetMAX™-Gold SIV Solutions—A USDA-licensed, real-time PCR screening and subtyping kits for the rapid detection of swine influenza virus (SIV)
  • SIV Detection Kit sensitivity of 98.4% and specificity of 99.1%
  • SIV Subtyping Detection Kit sensitivity of 98.2% and specificity of 100%
  • Real-time PCR technology helps improve reproducibility via assay standardization
  • Genomic targeting helps reduce risk of false negatives caused by genomic mutation
  • Kits are designed to provide rapid detection of SIV RNA extracted from porcine nasal swabs
  • USDA authorized representatives oversee the quality control process and the release of every batch
Find out more ›    Download product flyer ›