Ensure consistent material feed
Prevent bin overflows, plugged chutes, and empty bins with Thermo Scientific Ramsey level sensors. Our continuous level measurement devices penetrate dust, steam, vapors, and other harsh environmental obstacles to provide precise, reliable readings. Microwave or nuclear point level detection and control switches monitor the silo or bin level above the weighfeeder. We offer microwave-based, non-contact flow sensors for monitoring solid flows in pipelines, ducts, air slides, and at transfer points.
Help eliminate failure and maintenance issues with the Ramsey C-Level Continuous Level Indicator. This level measurement device is ideally suited for inventory monitoring and process control during the load-out or filling of bins and vessels containing bulk solids or liquids.
Prevent jitter and provide stability in your hazardous area applications with the Ramsey Mercury-Free Tilt Sensor. Various probe assemblies are available to suit a wide range of environmental conditions.
Invest in peace of mind with a Thermo Scientific service agreement. Ensure minimum downtime with known costs and priority response times. Let us tailor a plan to meet your needs. For parts, technical support, or field assistance, please call 800-445-3503.