Prevent accidents, protect equipment, and reduce shutdowns
We offer a variety of components for monitoring belt conveyors for bulk materials and other bulk process equipment. Thermo Scientific™ Ramsey™ conveyor safety switches and monitors analyze operating processes for potentially hazardous conditions and activate an alarm when they occur. This helps to keep your personnel safe, your equipment from being damaged, and your profits from becoming losses.
Prevent damage of expensive downstream equipment from tramp metal on conveyor belts. The Ramsey Oretronic IV Tramp Metal Detector can detect all types of metallic scrap, including bucket teeth, manganese steel mantles, bore crowns, bar scrap chains, tools, and more.
Monitor under speed or slippage conditions with the Ramsey Model 60-23P Under Speed Switch. This conveyor safety switch monitors the rotational velocity of conveyor belts, bucket elevators, and other types of rotating equipment.
Identify potentially hazardous situations with your with your bulk handling equipment with the Ramsey Belt Misalignment Switch. This conveyor safety switch monitors the position and tracking of conveyor belts and activates an alarm if the belt skews or misaligns.
Keep personnel safe and equipment from being damaged with this emergency shutdown device for conveyors or other equipment. The Ramsey Safety Cable Pull Switch is a conveyor safety switch that will activate an alarm when force is applied to the pull cable at any position.
Improve monitoring of underspeed, overspeed, and zero-speed conditions on bulk handling systems with the Ramsey Motion Monitor System.
Prevent your profit from turning into scrap
Millions of tires are discarded in landfills every year. Instead, they can be shredded to extract and recycle the steel they contain. Shredded tires are also used in construction, landscaping, and playground surfaces.
Because tires contain metal, the shredding process itself can damage costly shredding equipment. During the shredding process, a tramp metal detector provides an economical and reliable method of protecting expensive shredders, crushers, conveyors, and other equipment from potentially costly damage from wheel bolts, which reduces downtime and ensures the process stays operational. In general, tramp metal detectors are easy to install, operate, and calibrate, with no need for special tools or instruments.
Invest in peace of mind with a Thermo Scientific service agreement. Ensure minimum downtime with known costs and priority response times. Let us tailor a plan to meet your needs. For parts, technical support, or field assistance, please call 800-445-3503.