Thermo Scientific online analysis systems use prompt gamma neutron activation analysis (PGNAA) and pulsed fast thermal neutron activation (PFTNA) to determine the elemental composition of bulk raw materials. These analyzers are used in cement, coal, and sintering applications as well as other bulk materials such as copper, iron ore, phosphates, etc. to help ensure consistent material quality and process optimization. Use this comparison table to determine which PGNAA/PFTNA online analyzer is best for you.
Still have questions? We're here to help.
CB Omni Fusion | CQM Flex | ECA3 | |
![]() |
![]() |
![]() |
|
On belt analyzer | ✓ |
✓ |
|
Belt sizes (600 mm to 2200 mm) | ✓ |
✓ |
|
Sample stream analyzer | ✓ |
||
Elemental analysis | ✓ |
✓ |
✓ |
Coal | ✓ |
✓ |
|
Cement | ✓ |
||
Copper | ✓ |
||
Iron ore | ✓ |
||
Phosphates | ✓ |
||
Other bulk materials | ✓ |
||
Neutron generator capable | ✓ |
✓ |
|
Cf-252 capable | ✓ |
✓ |
✓ |