Assure certainty in routine results
Thermo Scientific Routine Vials and Caps are ideal for use in most standard HPLC and GC applications and high-throughput analyses. These quality autosampler vials and caps provide consistent results for your standard and high-throughput pharmaceutical, environmental, and food and beverage applications.
Delivering quality and performance
- Consistency at quantification levels
- Highly reproducible for samples >120µL
- Long-term sample stability for most analytes
- High solvent compatibility for mainstream HPLC and GC applications
- Compatible with standard HPLC and GC autosamplers
Quality autosampler vials in common types and sizes
- 1st hydrolytic class glass 51 expansion meets all current USP and Pharmacopeia criteria
- Short thread screw vials and caps: convenient handling of mid volatiles
- Crimp vials and caps: for low evaporation of highly volatile samples
- Snap vials and caps: convenient handling of low volatile samples
- Amber vials ensure integrity for light sensitive samples
Options for caps and septum for standard HPLC and GC requirements
- Septum have standard solvent stability
- Natural Rubber for GC applications & multiple injections
- Red silicone for GC and thin needles
- White high quality silicone for HPLC
- Bonded cap-septa/pre-slit septa to support instrumentation
Routine vials and caps
Routine screw top vials, caps, and septum
2mL glass screw vials, wide opening 12 x 32mm
Catalog | Description | Total volume | Usable volume | Residual volume | Pack of | |
---|---|---|---|---|---|---|
60180A-SV9-1 | Clear Glass Screw Vial, 9mm Short Thread | 2mL | 1.5mL | <120µL | 100 | Order |
60180A-SV9-1P | Clear Glass Screw Vial, 9mm Short Thread with ID Patch | 2mL | 1.5mL | <120µL | 100 | Order |
60180A-SV9-2P | Amber Glass Screw Vial, 9mm Short Thread with ID Patch | 2mL | 1.5mL | <120µL | 100 | Order |
9mm caps and septum for 2mL screw top vials above
Catalog | Description | Note | Hardness shore | Thickness | Pack of | |
---|---|---|---|---|---|---|
60180A-SC9NRT1 | 9mm Black Screw Cap, Natural Rubber/ PTFE Septa | Standard GC Septa | 60 | 1 | 100 | Order |
60180A-SC9ST1 | 9mm Black Screw Cap, Red Silicone/ Clear PTFE Septa | Standard HPLC Septa | 60 | 1 | 100 | Order |
60180A-SC9STS1 | 9mm Black Screw Cap, Red Silicone/ Clear PTFE Septa, Slitted | Standard HPLC Septa | 60 | 1 | 100 | Order |
60180A-SC9STB1 | 9mm Blue Screw Cap, Silicone/PTFE Bonded Septa | Standard HPLC Septa | 45 | 1 | 100 | Order |
60180A-SC9STBS1 | 9mm Blue Screw Cap, Silicone/PTFE Bonded Septa, Slitted | Special HPLC Septa | 45 | 1 | 100 | Order |
Routine crimp top vials, caps, and septum
2mL crimp top vials, wide opening
Catalog | Description | Total volume | Usable volume | Residual volume | Pack of | |
---|---|---|---|---|---|---|
60180A-CV11-1 | Clear Glass 11mm Crimp Top Vial | 2mL | 1.5mL | <120µL | 100 | Order |
60180A-CV11-1P | Clear Glass 11mm Crimp Top Vial with ID Patch | 2mL | 1.5mL | <120µL | 100 | Order |
60180A-CV11-2P | Amber Glass 11mm Crimp Top Vial with ID Patch | 2mL | 1.5mL | <120µL | 100 | Order |
Aluminum caps for 2mL crimp top vials above
Catalog | Description | Note | Hardness shore | Thickness | Pack of | |
---|---|---|---|---|---|---|
60180A-CC11NRT1 | 11mm Crimp Cap, Natural Rubber/ PTFE Septa | Standard GC Septa | 60 | 1 | 100 | Order |
60180A-CC11ST1 | 11mm Crimp Cap, Red Silicone/ Clear PTFE Septa | Standard HPLC Septa | 60 | 1 | 100 | Order |
Routine snap top vials, caps, and septum
2mL glass snap vials, wide opening 12 x 32mm
Catalog | Description | Total volume | Usable volume | Residual volume | Pack of | |
---|---|---|---|---|---|---|
60180A-RV11-1 | Clear Glass 11mm Snap Vial | 2mL | 1.5mL | <120µL | 100 | Order |
60180A-RV11-1P | Clear Glass 11mm Snap Vial with ID Patch | 2mL | 1.5mL | <120µL | 100 | Order |
60180A-RV11-2P | Amber Glass 11mm Snap Vial with ID Patch | 2mL | 1.5mL | <120µL | 100 | Order |
Snap caps for 2mL snap vials above
Catalog | Description | Septum | Hardness shore | Thickness | Pack of | |
---|---|---|---|---|---|---|
60180A-RC11NRT1 | 11mm Clear Snap Cap, with Natural Rubber/PTFE Septa | Standard GC Septa | 60 | 1 | 100 | Order |
60180A-RC11ST1 | 11mm Clear Snap Cap, Red Silicone/Clear PTFE Septa | Standard HPLC Septa | 60 | 1 | 100 | Order |
60180A-RC11STS1 | 11mm Clear Snap Cap, Red Silicone/Clear PTFE Septa, Slitted | Standard HPLC Septa | 60 | 1 | 100 | Order |
Routine screw top vials, caps, and septum
2mL glass screw vials, wide opening 12 x 32mm
Catalog | Description | Total volume | Usable volume | Residual volume | Pack of | |
---|---|---|---|---|---|---|
60180A-SV9-1 | Clear Glass Screw Vial, 9mm Short Thread | 2mL | 1.5mL | <120µL | 100 | Order |
60180A-SV9-1P | Clear Glass Screw Vial, 9mm Short Thread with ID Patch | 2mL | 1.5mL | <120µL | 100 | Order |
60180A-SV9-2P | Amber Glass Screw Vial, 9mm Short Thread with ID Patch | 2mL | 1.5mL | <120µL | 100 | Order |
9mm caps and septum for 2mL screw top vials above
Catalog | Description | Note | Hardness shore | Thickness | Pack of | |
---|---|---|---|---|---|---|
60180A-SC9NRT1 | 9mm Black Screw Cap, Natural Rubber/ PTFE Septa | Standard GC Septa | 60 | 1 | 100 | Order |
60180A-SC9ST1 | 9mm Black Screw Cap, Red Silicone/ Clear PTFE Septa | Standard HPLC Septa | 60 | 1 | 100 | Order |
60180A-SC9STS1 | 9mm Black Screw Cap, Red Silicone/ Clear PTFE Septa, Slitted | Standard HPLC Septa | 60 | 1 | 100 | Order |
60180A-SC9STB1 | 9mm Blue Screw Cap, Silicone/PTFE Bonded Septa | Standard HPLC Septa | 45 | 1 | 100 | Order |
60180A-SC9STBS1 | 9mm Blue Screw Cap, Silicone/PTFE Bonded Septa, Slitted | Special HPLC Septa | 45 | 1 | 100 | Order |
Routine crimp top vials, caps, and septum
2mL crimp top vials, wide opening
Catalog | Description | Total volume | Usable volume | Residual volume | Pack of | |
---|---|---|---|---|---|---|
60180A-CV11-1 | Clear Glass 11mm Crimp Top Vial | 2mL | 1.5mL | <120µL | 100 | Order |
60180A-CV11-1P | Clear Glass 11mm Crimp Top Vial with ID Patch | 2mL | 1.5mL | <120µL | 100 | Order |
60180A-CV11-2P | Amber Glass 11mm Crimp Top Vial with ID Patch | 2mL | 1.5mL | <120µL | 100 | Order |
Aluminum caps for 2mL crimp top vials above
Catalog | Description | Note | Hardness shore | Thickness | Pack of | |
---|---|---|---|---|---|---|
60180A-CC11NRT1 | 11mm Crimp Cap, Natural Rubber/ PTFE Septa | Standard GC Septa | 60 | 1 | 100 | Order |
60180A-CC11ST1 | 11mm Crimp Cap, Red Silicone/ Clear PTFE Septa | Standard HPLC Septa | 60 | 1 | 100 | Order |
Routine snap top vials, caps, and septum
2mL glass snap vials, wide opening 12 x 32mm
Catalog | Description | Total volume | Usable volume | Residual volume | Pack of | |
---|---|---|---|---|---|---|
60180A-RV11-1 | Clear Glass 11mm Snap Vial | 2mL | 1.5mL | <120µL | 100 | Order |
60180A-RV11-1P | Clear Glass 11mm Snap Vial with ID Patch | 2mL | 1.5mL | <120µL | 100 | Order |
60180A-RV11-2P | Amber Glass 11mm Snap Vial with ID Patch | 2mL | 1.5mL | <120µL | 100 | Order |
Snap caps for 2mL snap vials above
Catalog | Description | Septum | Hardness shore | Thickness | Pack of | |
---|---|---|---|---|---|---|
60180A-RC11NRT1 | 11mm Clear Snap Cap, with Natural Rubber/PTFE Septa | Standard GC Septa | 60 | 1 | 100 | Order |
60180A-RC11ST1 | 11mm Clear Snap Cap, Red Silicone/Clear PTFE Septa | Standard HPLC Septa | 60 | 1 | 100 | Order |
60180A-RC11STS1 | 11mm Clear Snap Cap, Red Silicone/Clear PTFE Septa, Slitted | Standard HPLC Septa | 60 | 1 | 100 | Order |
Not sure if you need Premium or Routine Autosampler Vials?
Premium vials and caps
- Autosampler Vials
- Autosampler Vial Closures and Septa
- Autosampler Vial Kits
- Certified Vial Kits
- Specialty Vials (EPA, sample storage, 4mL, headspace vials)
- Inert Vials and Inserts
- Autosampler Vial Inserts
Chromatography Consumables Resources and Information
Find interactive tools and calculators, guides to select the right products, equivalent product finders, and get in contact with our consumables technical support team to answer your application questions.
Workflow product selector
Select Chromatography Consumables and Columns for your Pharma, Biopharma, Clinical, Food & Beverage or Environmental workflow.
Resources
CMD SchemaApp code