Webinar: Workflows for robust and sensitive separation of mAbs, intact proteins at subunit and peptide level by LC-MS
Overview
Enhanced performance of low-flow LC-MS and HPLC separations
Low-flow chromatography is ideal when detailed sample information is required from small sample volumes, especially for bottom-up and top-down proteomics research. Compared with analytical flow LC-MS (>100 µL/min), the signal with low-flow (<100 µL/min) can be many times higher. Low-flow columns—also known as micro, nano, and capillary columns—have a smaller diameter compared to analytical columns.
Thermo Scientific nano-, capillary-, and micro-flow columns offer excellent sensitivity and resolution in easy-to-use formats:
- Thermo Scientific EASY-Spray HPLC Columns
- Thermo Scientific double nanoViper HPLC Columns
- Thermo Scientific µPAC HPLC Columns
Learn more about the power of low-flow columns from Thermo Fisher Scientific — technology that’s beyond brilliant.
Low-flow columns for reliable and robust data
Low-flow HPLC column formats | ||
EASY-Spray HPLC Columns | Double nanoViper HPLC Columns | µPAC HPLC Columns |
![]() | ![]() | ![]() |
EASY-Spray columns
![]() | |
Choose an EASY-Spray column when: | What makes an EASY-Spray column special? |
---|---|
| Unique design provides uncompromised performance in an ease of use format for nano and capillary LC-MS analysis. Features for optimum data quality:
|
Recommended EASY-Spray UHPLC columns by application
EASY-Spray PepMap Neo UHPLC columns
The Thermo Scientific EASY-Spray PepMap Neo UHPLC columns are perfect for bottom-up proteomics. Packed at higher pressure and rated to 1500 bar, they provide consistent column-to-column performance, long column lifetime, and excellent efficiency. These benefits are true at any pressure.
Catalog number | Product line | Particle size (µm) | Column ID (µm) | Column length (mm) | Order |
---|---|---|---|---|---|
ES75150PN | PepMap Neo columns | 2 | 75 | 150 | Order now › |
ES75500PN | PepMap Neo columns | 2 | 75 | 500 | Order now › |
ES75750PN | PepMap Neo columns | 2 | 75 | 750 | Order now › |
Also see: EASY-Spray PepMap HPLC columns
EASY-Spray MAbPac RP columns
The Thermo Scientific MAbPac RP capillary column is best suited for the characterization of intact proteins in top-down proteomics, clinical and anti-doping applications where sample amount is limited, or sensitivity is crucial.
Catalog number | Product line | Particle size (µm) | Column length (mm) | Column ID (µm) | Order |
---|---|---|---|---|---|
ES907 | MAbPac RP column | 4 | 150 | 150 | Order now › |
Traps and accessories
For the best performance from your EASY-Spray column, consider investing in the following accessories.
Traps
Catalog number | Product line | Particle size (µm) | Column ID (µm) | Media bed length (mm) | Trap length (mm) | Union type | Order |
---|---|---|---|---|---|---|---|
174500 | PepMap Neo Trap Cartridge 3PK | 5 | 300 | 5 | N/A | N/A | Order now › |
164199 | PepMap nanotrap 500 bar 2PK | 5 | 100 | 20 | 150 | Nut/sleeve | Order now › |
164750 | PepMap nanotrap 500 bar 2PK | 5 | 100 | 20 | 150 | Double nanoViper | Order now › |
164535 | PepMap nanotrap 500 bar 2PK | 3 | 75 | 20 | 150 | Double nanoViper | Order now › |
164946 | PepMap nanotrap 1200 bar 2PK | 3 | 75 | 20 | 70 | Double nanoViper | Order now › |
164213 | PepMap nanotrap 500 bar 2PK | 5 | 200 | 20 | 150 | Nut/sleeve | Order now › |
PEEK tubing and trap holder
Catalog number | Product line | For use with | Order |
---|---|---|---|
174501 | PEEK with nanoViper fittings 30 µm X100 mm 2PK 1500 bar | Low-flow PepMap columns | Order now › |
174502 | Trap holder + nanoViper fittings kit 1500 bar | Low-flow PepMap columns | Order now › |
Double nanoViper columns
![]() | |
Choose these columns when: | What makes these columns special? |
---|---|
| These stand-alone nano-, capillary, and micro-flow columns are:
|
Recommended nano- and capillary HPLC columns by application
Double nanoViper PepMap Neo UHPLC columns
Use these columns for peptide bottom-up proteomics studies. They are packed at higher pressure and rated a 1500 bar and provide consistent column-to-column performance, long column lifetime, and excellent efficiency. These benefits are true at any pressure.
Catalog number | Product line | Particle size (µm) | Column ID (µm) | Column length (mm) | Order |
---|---|---|---|---|---|
DNV75150PN | PepMap Neo column | 2 | 75 | 150 | Order now › |
DNV75500PN | PepMap Neo column | 2 | 75 | 500 | Order now › |
DNV75750PN | PepMap Neo column | 2 | 75 | 750 | Order now › |
Also available: Single nanoViper PepMap C18 HPLC columns
MAbPac capillary reversed phase (RP) HPLC column
The Thermo Scientific MAbPac RP capillary column is best suited for the characterization of intact proteins in top-down proteomics, clinical, and anti-doping applications where sample amount is limited, or sensitivity is crucial.
Catalog number | Product line | Particle size (µm) | Column ID (µm) | Column length (mm) | Order |
---|---|---|---|---|---|
164947 | MAbPac RP column | 4 | 150 | 150 | Order now › |
Accucore 150 Amide HILIC HPLC column
- Separate hydrophilic biomolecules in HILIC mode
- Excellent for released glycan separations
Catalog number | Product line | Particle size (µm) | Column ID (µm) | Column length (mm) | Order |
---|---|---|---|---|---|
16726-157569 | Amide HILIC column | 2.6 | 75 | 150 | Order now › |
These emitters, nanoViper tubing kits, and unions offer easy connection from your LC system to an EASY-Spray source.
Catalog number | Product line | For use with | Order |
---|---|---|---|
6040.2304 | Two Viper unions | Double nanoViper columns | Order now › |
6041.5260 | nanoViper tubing 20 µm x 550 mm | Double nanoViper columns | Order now › |
ES993 | Emitter: 10 µm I.D. | Double nanoViper columns | Order now › |
ES994 | Emitter: 15 µm I.D | Double nanoViper columns | Order now › |
For the best performance from your double nanoViper column consider investing in the following accessories.
Catalog number | Product line | Particle size (µm) | Column ID (µm) | Media bed length (mm) | Trap length (mm) | Union type | Order |
---|---|---|---|---|---|---|---|
174500 | PepMap Neo Trap Cartridge 3PK | 5 | 300 | 5 | N/A | N/A | Order now › |
164199 | PepMap nanotrap 500 bar 2PK | 5 | 100 | 20 | 150 | Nut/sleeve | Order now › |
164750 | PepMap nanotrap 500 bar 2PK | 5 | 100 | 20 | 150 | Double nanoViper | Order now › |
164535 | PepMap nanotrap 500 bar 2PK | 3 | 75 | 20 | 150 | Double nanoViper | Order now › |
164946 | PepMap nanotrap 1200 bar 2PK | 3 | 75 | 20 | 70 | Double nanoViper | Order now › |
164213 | PepMap nanotrap 500 bar 2PK | 5 | 200 | 20 | 150 | Nut/sleeve | Order now › |
Catalog number | Product line | For use with | Order |
---|---|---|---|
174501 | PEEK with nanoViper fittings 30 µm X100 mm 2PK 1500 bar | Low-flow PepMap Neo columns | Order now › |
174502 | Trap holder + nanoViper fittings kit 1500 bar | Low-flow PepMap Neo columns | Order now › |
µPAC HPLC columns
![]() | |
Choose a µPAC HPLC column when: | What makes µPAC HPLC columns special? |
---|---|
| The unique separation path provides:
|
These HPLC columns are ideally suited for bottom-up proteomics where resolution is critical to the success of the analysis. Our µPAC HPLC columns provide excellent column-to-column reproducibility and peak capacity at a wide range of flow rates.
Catalogue number | Product line | Pillar diameter (µm) | Interpillar distance (µm) | Column length (cm) | Flow rate range (µL/min) | Order |
---|---|---|---|---|---|---|
COL-nano050G1B | µPAC pillar array | 5 | 2.5 | 50 | 0.1 – 2.0 | Order now › |
COL-nano200G1B | µPAC pillar array | 5 | 2.5 | 200 | 0.1 - 1.0 | Order now › |
COL-cap050G1B | µPAC pillar array (capillary flow) | 5 | 2.5 | 50 | 1.0 - 15.0 | Order now › |
Catalogue number | Product line | Pillar diameter (µm) | Interpillar distance (µm) | Column length (cm) | Order |
---|---|---|---|---|---|
COL-trpnano16G1B2 (Duo-Pack) | µPAC pillar array | 5 | 2.5 | 1 | Order now › |
Catalogue number | Product line | For use with | Order |
---|---|---|---|
EMI-flexionB | μPAC Flex iON Connect ESI-MS: Interface to connect μPAC nanoLC columns to a Nanospray Flex Series ion source | μPAC nano columns | Order now › |
EMI-easysprayB | μPAC Compatible EASY-Spray Emitter: Pre-assembled EASY-Spray emitter, grounding union, and transfer line to connect μPAC nanoLC columns to an EASY-Spray source | μPAC nano columns | Order now › |
Thermo Scientific Vanquish Neo UHPLC System
The all-in-one nano-, capillary-, and micro-flow LC system for high sensitivity LCMS workflows.
Low-flow HPLC column videos
Related products
Thermo Scientific SMART Digest Kits for Protein Digestion
Low-flow Columns Catalog
Low-flow chromatography consumables reference guide for LC-MS proteomics research
Download this guide and learn more about the portfolio of Thermo Scientific consumables for LC-MS proteomics research and the value they can bring to your lab.
Resources for low-flow HPLC columns
Brochures
Flyers
Application Notes
Best Practice Guides
Technical Notes
Chromatography consumables on-demand webinar series
Discover live and on-demand webinars that enable you to improve productivity, maintain compliance, and learn about the constantly evolving scientific landscape.