Concentrate samples for lower detection limits and increased sensitivity
Used primarily for high-purity water analysis, Thermo Scientific™ IonPac™ and IonSwift™ concentrator columns retain ions from a measured volume of aqueous sample matrix, thereby concentrating the analyte species and lowering detection limits. Concentrator columns can increase sensitivity by two to five orders of magnitude when compared to standard sample loops.
IC anion concentrator columns
Dionex™ AC-ER Anion Concentrator for RFIC-ER | Use this general purpose, low-dead-volume anion concentrator column to maximize the number of injections between eluent changes when using an RFIC-ER™ system. Use for a host of applications on RFIC-ER systems, including analysis of bromate to 5 ppb in drinking water, and analysis of common anions in ethanol. |
Dionex™ AMC-1 Anion Micro Concentrator Column | Reduce detection limits of inorganic and low molecular weight organic anions in ultrapure water by 2–5 orders of magnitude; novel solvent-compatible resin technology improves anion determinations for trace anions in the power generation and semiconductor industries. |
Dionex™ IonPac™ AC10 Concentrator Column | Use this concentrator with the Dionex™ IonPac™ AS10 Hydroxide-Selective Anion-Exchange Column to lower detection limits in high-purity water analysis |
Dionex™ IonPac™ AC15 Concentrator Column | Use this concentrator with the Dionex™ IonPac™ AS15 Hydroxide-Selective Anion-Exchange Column for ultratrace (part per trillion) analyses. |
Dionex™ IonPac™ Cryptand C1 Concentrator Column | Use this adjustable-capacity anion concentrator column for the determination of trace perchlorate in drinking and high-ionic-strength waters as specified in U.S. EPA Method 314.1. |
Dionex™ IonPac™ Low Pressure TAC Trace Anion Concentrator Columns (TAC-LP1, TAC-ULP1) | Use these pellicular low and ultralow pressure columns for general purpose anion concentration in high-purity water matrices for trace level analysis. Columns support carbonate/bicarbonate, borate, and hydroxide eluents. |
Dionex™ IonPac™ TAC-2 Trace Anion Concentrator Columns | Use this general purpose pellicular anion concentrator column for high-purity water analysis when using carbonate eluents. |
Dionex™ IonPac™ UTAC-1 Series Ultratrace Anion Concentrator Columns, (UTAC-LP1, UTAC-ULP1, UTAC-XLP1) | Choose from low, ultralow, and extremely low pressure formats to concentrate anions for trace-level analysis of high-purity water. |
Dionex™ IonPac™ UTAC-2 Series Ultratrace Anion Concentrator Columns, (UTAC-LP2, UTAC-ULP2, UTAC-XLP2) | Use these columns, available in low, ultralow, and extremely low pressure formats with improved backpressures, to concentrate anions for trace-level analysis of high-purity water. Compatible with carbonate/bicarbonate, borate, or hydroxide eluents with or without solvents. |
Dionex™ IonSwift™ MAC Monolith Anion Concentrator Columns (MAC-100, MAC-200) | Concentrate anions for general purpose or 2D-IC water analyses using these ultraclean, ultralow-backpressure, low-void volume monolithic concentrator columns. |
IC cation concentrator columns
Dionex™ IonPac™ Cation Eluent Purification Column (CEP-ER1) | Use this column with eluent regeneration kits. |
Dionex™ IonPac™ TCC Trace Cation Concentrator Columns | Preconcentrate cations for high-purity water analysis or trace analysis of transition metals with this pellicular cation-exchange, surface-sulfonated concentrator column. |
Dionex™ IonPac™ TCC Trace Cation Concentrators (TCC-LP1, TCC-ULP1, TCC-XLP1) | Choose from low, ultralow and extremely low pressure formats to concentrate cations for trace-level analysis of high-purity water. Recommended for use with carboxylated columns and monovalent eluents such as sulfuric acid, sulfonates, and hydrochloria acid. |
Dionex™ IonSwift™ MCC-100 Monolith Cation Concentrator Columns | Concentrate cations for general purpose high-purity water analyses and for trace ammonium and amines in high salt matrices by 2D-IC using ultralow pressure concentrator… |
Dionex™ MetPac™ CC-1 Cation Concentrator | Use this column in chelation IC and IC-ICP methods to concentrate cationic transition metals and lanthanide elements from high-ionic-strength matrices. |
Dionex™ TMC-1 Trace Metal Concentrator Column | Concentrate trace metals for chelation IC using the Dionex TMC-1 Trace Metal Concentrator Column. |
IC anion concentrator columns
Dionex™ AC-ER Anion Concentrator for RFIC-ER | Use this general purpose, low-dead-volume anion concentrator column to maximize the number of injections between eluent changes when using an RFIC-ER™ system. Use for a host of applications on RFIC-ER systems, including analysis of bromate to 5 ppb in drinking water, and analysis of common anions in ethanol. |
Dionex™ AMC-1 Anion Micro Concentrator Column | Reduce detection limits of inorganic and low molecular weight organic anions in ultrapure water by 2–5 orders of magnitude; novel solvent-compatible resin technology improves anion determinations for trace anions in the power generation and semiconductor industries. |
Dionex™ IonPac™ AC10 Concentrator Column | Use this concentrator with the Dionex™ IonPac™ AS10 Hydroxide-Selective Anion-Exchange Column to lower detection limits in high-purity water analysis |
Dionex™ IonPac™ AC15 Concentrator Column | Use this concentrator with the Dionex™ IonPac™ AS15 Hydroxide-Selective Anion-Exchange Column for ultratrace (part per trillion) analyses. |
Dionex™ IonPac™ Cryptand C1 Concentrator Column | Use this adjustable-capacity anion concentrator column for the determination of trace perchlorate in drinking and high-ionic-strength waters as specified in U.S. EPA Method 314.1. |
Dionex™ IonPac™ Low Pressure TAC Trace Anion Concentrator Columns (TAC-LP1, TAC-ULP1) | Use these pellicular low and ultralow pressure columns for general purpose anion concentration in high-purity water matrices for trace level analysis. Columns support carbonate/bicarbonate, borate, and hydroxide eluents. |
Dionex™ IonPac™ TAC-2 Trace Anion Concentrator Columns | Use this general purpose pellicular anion concentrator column for high-purity water analysis when using carbonate eluents. |
Dionex™ IonPac™ UTAC-1 Series Ultratrace Anion Concentrator Columns, (UTAC-LP1, UTAC-ULP1, UTAC-XLP1) | Choose from low, ultralow, and extremely low pressure formats to concentrate anions for trace-level analysis of high-purity water. |
Dionex™ IonPac™ UTAC-2 Series Ultratrace Anion Concentrator Columns, (UTAC-LP2, UTAC-ULP2, UTAC-XLP2) | Use these columns, available in low, ultralow, and extremely low pressure formats with improved backpressures, to concentrate anions for trace-level analysis of high-purity water. Compatible with carbonate/bicarbonate, borate, or hydroxide eluents with or without solvents. |
Dionex™ IonSwift™ MAC Monolith Anion Concentrator Columns (MAC-100, MAC-200) | Concentrate anions for general purpose or 2D-IC water analyses using these ultraclean, ultralow-backpressure, low-void volume monolithic concentrator columns. |
IC cation concentrator columns
Dionex™ IonPac™ Cation Eluent Purification Column (CEP-ER1) | Use this column with eluent regeneration kits. |
Dionex™ IonPac™ TCC Trace Cation Concentrator Columns | Preconcentrate cations for high-purity water analysis or trace analysis of transition metals with this pellicular cation-exchange, surface-sulfonated concentrator column. |
Dionex™ IonPac™ TCC Trace Cation Concentrators (TCC-LP1, TCC-ULP1, TCC-XLP1) | Choose from low, ultralow and extremely low pressure formats to concentrate cations for trace-level analysis of high-purity water. Recommended for use with carboxylated columns and monovalent eluents such as sulfuric acid, sulfonates, and hydrochloria acid. |
Dionex™ IonSwift™ MCC-100 Monolith Cation Concentrator Columns | Concentrate cations for general purpose high-purity water analyses and for trace ammonium and amines in high salt matrices by 2D-IC using ultralow pressure concentrator… |
Dionex™ MetPac™ CC-1 Cation Concentrator | Use this column in chelation IC and IC-ICP methods to concentrate cationic transition metals and lanthanide elements from high-ionic-strength matrices. |
Dionex™ TMC-1 Trace Metal Concentrator Column | Concentrate trace metals for chelation IC using the Dionex TMC-1 Trace Metal Concentrator Column. |
Dionex concentrator and trap columns data sheet
Find out more about our:
- Concentrator columns, which allow anion, cation, and chelation IC determinations at μg/L (ppb) and sub-μg/L concentrations
- Trap columns, which prevent contaminant interference during gradient IC